Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88908605-88908848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAMBPL1
Entrez Gene ID
57559
Gene Name
STAM binding protein like 1
Gene Aliases
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2
UniGene
Hs.16229
Gene Chromosome Location
Chr.10: 88879368-88923502 on Build GRCh38
Transcripts
NM_020799.3 XM_006717928.2 XM_006717930.3 XM_011539983.1 XM_011539985.1 XM_017016453.1 XM_017016454.1 XM_017016455.1 XM_017016456.1 XM_017016457.1 XM_017016458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4573873 c.272A>C p.N91T Chr.10: 88908725-88908725 on Build GRCh38
4851722 c.321G>C p.M107I Chr.10: 88908774-88908774 on Build GRCh38
267518 c.293G>A p.C98Y Chr.10: 88908746-88908746 on Build GRCh38
267519 c.293G>A p.C98Y Chr.10: 88908746-88908746 on Build GRCh38
4851721 c.321G>C p.M107I Chr.10: 88908774-88908774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777374811 Chr.10: 88908695-88908695 on Build GRCh38
rs753438982 Chr.10: 88908705-88908705 on Build GRCh38
rs759304206 Chr.10: 88908656-88908656 on Build GRCh38
rs963693714 Chr.10: 88908774-88908774 on Build GRCh38
rs767228192 Chr.10: 88908767-88908767 on Build GRCh38
rs573165834 Chr.10: 88908786-88908786 on Build GRCh38
rs755591685 Chr.10: 88908686-88908686 on Build GRCh38
rs766623833 Chr.10: 88908654-88908654 on Build GRCh38
rs767850094 Chr.10: 88908757-88908758 on Build GRCh38
rs753482836 Chr.10: 88908725-88908725 on Build GRCh38
rs764176508 Chr.10: 88908801-88908801 on Build GRCh38
rs755304246 Chr.10: 88908730-88908730 on Build GRCh38
rs756949249 Chr.10: 88908707-88908707 on Build GRCh38
rs368542029 Chr.10: 88908754-88908754 on Build GRCh38
rs751330354 Chr.10: 88908655-88908655 on Build GRCh38
rs950895608 Chr.10: 88908660-88908660 on Build GRCh38
rs750993477 Chr.10: 88908788-88908788 on Build GRCh38
rs773241470 Chr.10: 88908745-88908745 on Build GRCh38
rs771126052 Chr.10: 88908750-88908750 on Build GRCh38
rs935666100 Chr.10: 88908736-88908736 on Build GRCh38
rs867249937 Chr.10: 88908757-88908757 on Build GRCh38
rs760561106 Chr.10: 88908798-88908798 on Build GRCh38
rs371808752 Chr.10: 88908782-88908782 on Build GRCh38
rs767483215 Chr.10: 88908661-88908661 on Build GRCh38
rs181446155 Chr.10: 88908731-88908731 on Build GRCh38
rs769882182 Chr.10: 88908741-88908741 on Build GRCh38
rs58713776 Chr.10: 88908685-88908685 on Build GRCh38
rs577091365 Chr.10: 88908664-88908664 on Build GRCh38
rs774620760 Chr.10: 88908751-88908751 on Build GRCh38
rs753385909 Chr.10: 88908814-88908814 on Build GRCh38

View Full Product Details