Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76003008-76003233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK3
Entrez Gene ID
1018
Gene Name
cyclin dependent kinase 3
Gene Aliases
-
UniGene
Hs.706766
Gene Chromosome Location
Chr.17: 76000906-76005999 on Build GRCh38
Transcripts
NM_001258.2 NR_037709.1

Gene Symbol
TEN1-CDK3
Entrez Gene ID
100529145
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
Gene Aliases
C17orf106-CDK3
UniGene
Hs.706766
Gene Chromosome Location
Chr.17: 75979217-76005999 on Build GRCh38
Transcripts
NM_001258.2 NR_037709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984543 c.599A>C p.K200T Chr.17: 76003205-76003205 on Build GRCh38
3680561 c.610C>T p.P204S Chr.17: 76003216-76003216 on Build GRCh38
6625514 c.596G>A p.R199Q Chr.17: 76003202-76003202 on Build GRCh38
984542 c.598A>C p.K200Q Chr.17: 76003204-76003204 on Build GRCh38
3731816 c.618C>T p.D206D Chr.17: 76003224-76003224 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755363630 Chr.17: 76003173-76003175 on Build GRCh38
rs1031812965 Chr.17: 76003151-76003151 on Build GRCh38
rs998656939 Chr.17: 76003125-76003125 on Build GRCh38
rs971231359 Chr.17: 76003122-76003122 on Build GRCh38
rs757875069 Chr.17: 76003170-76003170 on Build GRCh38
rs910252409 Chr.17: 76003187-76003187 on Build GRCh38
rs200505631 Chr.17: 76003201-76003201 on Build GRCh38
rs779790633 Chr.17: 76003208-76003208 on Build GRCh38
rs959111223 Chr.17: 76003076-76003076 on Build GRCh38
rs1005826277 Chr.17: 76003054-76003054 on Build GRCh38
rs758771724 Chr.17: 76003202-76003202 on Build GRCh38
rs371131209 Chr.17: 76003159-76003159 on Build GRCh38
rs1033336140 Chr.17: 76003057-76003057 on Build GRCh38
rs1013248047 Chr.17: 76003121-76003121 on Build GRCh38
rs200300738 Chr.17: 76003165-76003165 on Build GRCh38
rs1025150735 Chr.17: 76003136-76003136 on Build GRCh38
rs752793978 Chr.17: 76003161-76003161 on Build GRCh38
rs192675337 Chr.17: 76003140-76003140 on Build GRCh38
rs554150186 Chr.17: 76003162-76003162 on Build GRCh38
rs1012827873 Chr.17: 76003080-76003080 on Build GRCh38
rs888716709 Chr.17: 76003052-76003052 on Build GRCh38
rs779552695 Chr.17: 76003183-76003183 on Build GRCh38
rs1024071506 Chr.17: 76003083-76003083 on Build GRCh38

View Full Product Details