Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72294323-72294596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPB
Entrez Gene ID
81570
Gene Name
ClpB homolog, mitochondrial AAA ATPase chaperonin
Gene Aliases
ANKCLB, HSP78, MEGCANN, MGCA7, SKD3
UniGene
Hs.523877
Gene Chromosome Location
Chr.11: 72292425-72434684 on Build GRCh38
Transcripts
NM_001258392.2 NM_001258393.2 NM_001258394.2 NM_030813.5 XM_005274320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5907559 c.1666G>A p.D556N Chr.11: 72294429-72294429 on Build GRCh38
1723054 c.1770+1G>C p.? Chr.11: 72294324-72294324 on Build GRCh38
3453005 c.1695C>T p.I565I Chr.11: 72294400-72294400 on Build GRCh38
5704174 c.1715G>T p.C572F Chr.11: 72294380-72294380 on Build GRCh38
7162329 c.1662G>A p.R554R Chr.11: 72294433-72294433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983210578 Chr.11: 72294446-72294446 on Build GRCh38
rs876657402 Chr.11: 72294410-72294410 on Build GRCh38
rs374032537 Chr.11: 72294357-72294357 on Build GRCh38
rs764873441 Chr.11: 72294418-72294418 on Build GRCh38
rs758906536 Chr.11: 72294400-72294400 on Build GRCh38
rs772629590 Chr.11: 72294489-72294489 on Build GRCh38
rs748619997 Chr.11: 72294476-72294476 on Build GRCh38
rs768632963 Chr.11: 72294415-72294416 on Build GRCh38
rs747281970 Chr.11: 72294492-72294492 on Build GRCh38
rs765975867 Chr.11: 72294386-72294386 on Build GRCh38
rs754999129 Chr.11: 72294462-72294462 on Build GRCh38
rs768787799 Chr.11: 72294349-72294349 on Build GRCh38
rs879014880 Chr.11: 72294567-72294567 on Build GRCh38
rs759256728 Chr.11: 72294573-72294573 on Build GRCh38
rs150857620 Chr.11: 72294395-72294395 on Build GRCh38
rs570946878 Chr.11: 72294457-72294457 on Build GRCh38
rs200970392 Chr.11: 72294385-72294385 on Build GRCh38
rs776265668 Chr.11: 72294467-72294468 on Build GRCh38
rs576652298 Chr.11: 72294434-72294434 on Build GRCh38
rs752634197 Chr.11: 72294424-72294424 on Build GRCh38
rs780073036 Chr.11: 72294450-72294450 on Build GRCh38
rs556496847 Chr.11: 72294544-72294544 on Build GRCh38
rs764818576 Chr.11: 72294576-72294576 on Build GRCh38
rs749270611 Chr.11: 72294345-72294345 on Build GRCh38
rs758459970 Chr.11: 72294429-72294429 on Build GRCh38
rs376110052 Chr.11: 72294358-72294358 on Build GRCh38
rs200299650 Chr.11: 72294435-72294435 on Build GRCh38
rs981754949 Chr.11: 72294412-72294412 on Build GRCh38
rs772407202 Chr.11: 72294384-72294384 on Build GRCh38
rs942081291 Chr.11: 72294515-72294515 on Build GRCh38
rs757088240 Chr.11: 72294447-72294447 on Build GRCh38
rs773556958 Chr.11: 72294490-72294490 on Build GRCh38
rs771333751 Chr.11: 72294570-72294570 on Build GRCh38
rs1011708876 Chr.11: 72294543-72294543 on Build GRCh38
rs559051587 Chr.11: 72294453-72294453 on Build GRCh38
rs776356646 Chr.11: 72294399-72294399 on Build GRCh38
rs779010391 Chr.11: 72294463-72294463 on Build GRCh38
rs537999414 Chr.11: 72294535-72294535 on Build GRCh38

View Full Product Details