Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95943316-95943543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
UniGene
Hs.424907
Gene Chromosome Location
Chr.12: 95943293-95968592 on Build GRCh38
Transcripts
NM_152435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749235 c.49C>T p.L17L Chr.12: 95943447-95943447 on Build GRCh38
4792830 c.126G>C p.L42L Chr.12: 95943524-95943524 on Build GRCh38
6426483 c.4G>A p.A2T Chr.12: 95943402-95943402 on Build GRCh38
7280248 c.110T>C p.L37P Chr.12: 95943508-95943508 on Build GRCh38
3749234 c.7A>G p.S3G Chr.12: 95943405-95943405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376246479 Chr.12: 95943474-95943474 on Build GRCh38
rs965697187 Chr.12: 95943457-95943457 on Build GRCh38
rs7955759 Chr.12: 95943386-95943386 on Build GRCh38
rs1023682827 Chr.12: 95943388-95943388 on Build GRCh38
rs1436121 Chr.12: 95943447-95943447 on Build GRCh38
rs764568171 Chr.12: 95943383-95943383 on Build GRCh38
rs750622921 Chr.12: 95943401-95943401 on Build GRCh38
rs766702790 Chr.12: 95943407-95943407 on Build GRCh38
rs934128292 Chr.12: 95943493-95943493 on Build GRCh38
rs920961016 Chr.12: 95943494-95943494 on Build GRCh38
rs878856453 Chr.12: 95943464-95943464 on Build GRCh38
rs576513008 Chr.12: 95943389-95943389 on Build GRCh38
rs537265116 Chr.12: 95943390-95943390 on Build GRCh38
rs896743060 Chr.12: 95943351-95943351 on Build GRCh38
rs373119843 Chr.12: 95943352-95943352 on Build GRCh38
rs975522989 Chr.12: 95943463-95943463 on Build GRCh38
rs17852641 Chr.12: 95943521-95943521 on Build GRCh38
rs7955450 Chr.12: 95943405-95943405 on Build GRCh38
rs777609579 Chr.12: 95943461-95943461 on Build GRCh38
rs757021541 Chr.12: 95943501-95943501 on Build GRCh38
rs539199262 Chr.12: 95943375-95943375 on Build GRCh38
rs909871561 Chr.12: 95943444-95943444 on Build GRCh38
rs956790305 Chr.12: 95943396-95943396 on Build GRCh38
rs921468506 Chr.12: 95943468-95943468 on Build GRCh38
rs749204970 Chr.12: 95943462-95943462 on Build GRCh38
rs751745578 Chr.12: 95943410-95943410 on Build GRCh38

View Full Product Details