Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57766814-57767073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27B1
Entrez Gene ID
1594
Gene Name
cytochrome P450 family 27 subfamily B member 1
Gene Aliases
CP2B, CYP1, CYP1alpha, CYP27B, P450c1, PDDR, VDD1, VDDR, VDDRI, VDR
UniGene
Hs.524528
Gene Chromosome Location
Chr.12: 57762334-57767193 on Build GRCh38
Transcripts
NM_000785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1989877 c.171delG p.L58fs*20 Chr.12: 57766871-57766871 on Build GRCh38
1989876 c.189G>T p.E63D Chr.12: 57766853-57766853 on Build GRCh38
6342273 c.37C>A p.H13N Chr.12: 57767005-57767005 on Build GRCh38
7251383 c.14T>C p.L5P Chr.12: 57767028-57767028 on Build GRCh38
1363438 c.171_172insG p.L58fs*275 Chr.12: 57766870-57766871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368820787 Chr.12: 57766960-57766960 on Build GRCh38
rs748991170 Chr.12: 57766966-57766966 on Build GRCh38
rs201343041 Chr.12: 57766897-57766897 on Build GRCh38
rs577501884 Chr.12: 57767020-57767020 on Build GRCh38
rs141890302 Chr.12: 57766940-57766940 on Build GRCh38
rs778438734 Chr.12: 57767021-57767021 on Build GRCh38
rs772434439 Chr.12: 57766868-57766868 on Build GRCh38
rs202032208 Chr.12: 57766876-57766876 on Build GRCh38
rs752594802 Chr.12: 57766912-57766912 on Build GRCh38
rs755512223 Chr.12: 57767011-57767011 on Build GRCh38
rs868704228 Chr.12: 57766847-57766847 on Build GRCh38
rs769487954 Chr.12: 57766961-57766961 on Build GRCh38
rs761803324 Chr.12: 57767056-57767056 on Build GRCh38
rs955639760 Chr.12: 57766942-57766942 on Build GRCh38
rs374801900 Chr.12: 57767025-57767025 on Build GRCh38
rs768601263 Chr.12: 57767051-57767051 on Build GRCh38
rs776510825 Chr.12: 57767053-57767053 on Build GRCh38
rs771825668 Chr.12: 57767036-57767036 on Build GRCh38
rs765408119 Chr.12: 57766995-57766995 on Build GRCh38
rs757388190 Chr.12: 57766941-57766941 on Build GRCh38
rs537715358 Chr.12: 57766988-57766988 on Build GRCh38
rs775231921 Chr.12: 57767046-57767046 on Build GRCh38
rs911184682 Chr.12: 57766965-57766965 on Build GRCh38
rs772736324 Chr.12: 57766904-57766904 on Build GRCh38
rs372223837 Chr.12: 57767001-57767001 on Build GRCh38
rs755845434 Chr.12: 57767037-57767038 on Build GRCh38
rs780429096 Chr.12: 57766959-57766959 on Build GRCh38
rs927941015 Chr.12: 57766934-57766934 on Build GRCh38
rs376304260 Chr.12: 57766872-57766872 on Build GRCh38
rs757673918 Chr.12: 57766858-57766858 on Build GRCh38
rs202028173 Chr.12: 57766999-57766999 on Build GRCh38
rs760753732 Chr.12: 57766919-57766919 on Build GRCh38
rs764141450 Chr.12: 57766925-57766925 on Build GRCh38
rs748833551 Chr.12: 57767016-57767016 on Build GRCh38
rs779382219 Chr.12: 57766943-57766943 on Build GRCh38
rs770470667 Chr.12: 57766967-57766967 on Build GRCh38
rs762682469 Chr.12: 57766878-57766878 on Build GRCh38
rs745516871 Chr.12: 57767028-57767028 on Build GRCh38
rs758703209 Chr.12: 57767000-57767000 on Build GRCh38
rs758749494 Chr.12: 57766954-57766954 on Build GRCh38
rs770828560 Chr.12: 57766886-57766886 on Build GRCh38
rs764196493 Chr.12: 57766986-57766986 on Build GRCh38
rs753998021 Chr.12: 57766838-57766838 on Build GRCh38
rs1050216132 Chr.12: 57767017-57767017 on Build GRCh38
rs767074885 Chr.12: 57766871-57766871 on Build GRCh38
rs763437121 Chr.12: 57766870-57766871 on Build GRCh38
rs777419232 Chr.12: 57766964-57766964 on Build GRCh38
rs774009424 Chr.12: 57766971-57766971 on Build GRCh38
rs761902845 Chr.12: 57766993-57766993 on Build GRCh38
rs754122776 Chr.12: 57766939-57766939 on Build GRCh38
rs771838478 Chr.12: 57766982-57766982 on Build GRCh38
rs759569512 Chr.12: 57766894-57766894 on Build GRCh38
rs769363080 Chr.12: 57766875-57766875 on Build GRCh38
rs750773217 Chr.12: 57766952-57766952 on Build GRCh38
rs777477723 Chr.12: 57767012-57767012 on Build GRCh38
rs964139740 Chr.12: 57766985-57766985 on Build GRCh38
rs139558307 Chr.12: 57766892-57766892 on Build GRCh38
rs775461299 Chr.12: 57766984-57766984 on Build GRCh38
rs746424645 Chr.12: 57766865-57766865 on Build GRCh38
rs752330549 Chr.12: 57766999-57767000 on Build GRCh38
rs576826652 Chr.12: 57766853-57766853 on Build GRCh38
rs759285613 Chr.12: 57766976-57766976 on Build GRCh38
rs779138180 Chr.12: 57766860-57766860 on Build GRCh38
rs982250460 Chr.12: 57766920-57766920 on Build GRCh38
rs888983913 Chr.12: 57767037-57767037 on Build GRCh38
rs150648140 Chr.12: 57767002-57767002 on Build GRCh38

View Full Product Details