Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10138226-10138479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT1
Entrez Gene ID
1786
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
UniGene
Hs.202672
Gene Chromosome Location
Chr.19: 10133344-10195135 on Build GRCh38
Transcripts
NM_001130823.2 NM_001318730.1 NM_001318731.1 NM_001379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438455 c.4036G>C p.D1346H Chr.19: 10138470-10138470 on Build GRCh38
7001018 c.4115+8_4115+9GC>AT p.? Chr.19: 10138430-10138431 on Build GRCh38
6962692 c.4066A>G p.R1356G Chr.19: 10138440-10138440 on Build GRCh38
239627 c.4039G>A p.D1347N Chr.19: 10138467-10138467 on Build GRCh38
7234285 c.4104C>G p.S1368R Chr.19: 10138450-10138450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751166097 Chr.19: 10138396-10138396 on Build GRCh38
rs771522177 Chr.19: 10138409-10138409 on Build GRCh38
rs987619801 Chr.19: 10138268-10138268 on Build GRCh38
rs775701177 Chr.19: 10138423-10138423 on Build GRCh38
rs906682113 Chr.19: 10138376-10138376 on Build GRCh38
rs898269491 Chr.19: 10138315-10138329 on Build GRCh38
rs372346762 Chr.19: 10138398-10138398 on Build GRCh38
rs894060134 Chr.19: 10138417-10138417 on Build GRCh38
rs931538574 Chr.19: 10138383-10138383 on Build GRCh38
rs772278199 Chr.19: 10138418-10138418 on Build GRCh38
rs941042817 Chr.19: 10138315-10138315 on Build GRCh38
rs536273688 Chr.19: 10138312-10138312 on Build GRCh38
rs372158548 Chr.19: 10138377-10138377 on Build GRCh38
rs756058705 Chr.19: 10138409-10138410 on Build GRCh38
rs975674683 Chr.19: 10138358-10138358 on Build GRCh38
rs369000824 Chr.19: 10138426-10138426 on Build GRCh38
rs982660129 Chr.19: 10138307-10138307 on Build GRCh38
rs1047018976 Chr.19: 10138355-10138355 on Build GRCh38
rs565076498 Chr.19: 10138410-10138410 on Build GRCh38
rs951297564 Chr.19: 10138302-10138302 on Build GRCh38
rs572044408 Chr.19: 10138267-10138267 on Build GRCh38
rs576420043 Chr.19: 10138327-10138327 on Build GRCh38
rs867377260 Chr.19: 10138270-10138270 on Build GRCh38
rs548479350 Chr.19: 10138292-10138299 on Build GRCh38
rs955844747 Chr.19: 10138262-10138262 on Build GRCh38
rs1057452419 Chr.19: 10138425-10138425 on Build GRCh38
rs543712087 Chr.19: 10138404-10138404 on Build GRCh38
rs779688616 Chr.19: 10138391-10138391 on Build GRCh38
rs189497536 Chr.19: 10138316-10138316 on Build GRCh38
rs1016656594 Chr.19: 10138302-10138316 on Build GRCh38
rs375293718 Chr.19: 10138412-10138412 on Build GRCh38
rs977021400 Chr.19: 10138294-10138294 on Build GRCh38
rs949809894 Chr.19: 10138346-10138347 on Build GRCh38
rs754022171 Chr.19: 10138414-10138414 on Build GRCh38
rs776250179 Chr.19: 10138439-10138439 on Build GRCh38
rs201167482 Chr.19: 10138446-10138446 on Build GRCh38
rs369991149 Chr.19: 10138403-10138403 on Build GRCh38

View Full Product Details