Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58840593-58840848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs145893409 Chr.20: 58840724-58840724 on Build GRCh38
rs74589442 Chr.20: 58840670-58840670 on Build GRCh38
rs772141516 Chr.20: 58840738-58840740 on Build GRCh38
rs142690161 Chr.20: 58840753-58840753 on Build GRCh38
rs758925149 Chr.20: 58840694-58840694 on Build GRCh38
rs756503903 Chr.20: 58840637-58840637 on Build GRCh38
rs759229542 Chr.20: 58840784-58840784 on Build GRCh38
rs765883995 Chr.20: 58840683-58840683 on Build GRCh38
rs369158171 Chr.20: 58840651-58840651 on Build GRCh38
rs1032026231 Chr.20: 58840806-58840806 on Build GRCh38
rs919438504 Chr.20: 58840692-58840692 on Build GRCh38
rs749569559 Chr.20: 58840642-58840642 on Build GRCh38
rs79709641 Chr.20: 58840786-58840786 on Build GRCh38
rs778945290 Chr.20: 58840739-58840739 on Build GRCh38
rs773839807 Chr.20: 58840779-58840779 on Build GRCh38
rs374954928 Chr.20: 58840639-58840639 on Build GRCh38
rs773279644 Chr.20: 58840795-58840795 on Build GRCh38
rs371055001 Chr.20: 58840698-58840698 on Build GRCh38
rs773930351 Chr.20: 58840685-58840685 on Build GRCh38
rs758514619 Chr.20: 58840748-58840748 on Build GRCh38
rs769719956 Chr.20: 58840756-58840756 on Build GRCh38
rs756663141 Chr.20: 58840718-58840718 on Build GRCh38
rs748047944 Chr.20: 58840659-58840659 on Build GRCh38
rs745982259 Chr.20: 58840745-58840745 on Build GRCh38
rs771753113 Chr.20: 58840796-58840796 on Build GRCh38
rs938067305 Chr.20: 58840785-58840785 on Build GRCh38
rs770021477 Chr.20: 58840639-58840640 on Build GRCh38
rs762734492 Chr.20: 58840677-58840677 on Build GRCh38
rs980818308 Chr.20: 58840773-58840773 on Build GRCh38
rs76928384 Chr.20: 58840751-58840751 on Build GRCh38
rs761327919 Chr.20: 58840807-58840807 on Build GRCh38
rs181594534 Chr.20: 58840643-58840643 on Build GRCh38
rs775124978 Chr.20: 58840800-58840800 on Build GRCh38
rs898544242 Chr.20: 58840617-58840617 on Build GRCh38
rs112322423 Chr.20: 58840826-58840826 on Build GRCh38
rs774037468 Chr.20: 58840653-58840655 on Build GRCh38
rs764608006 Chr.20: 58840719-58840719 on Build GRCh38
rs754145936 Chr.20: 58840721-58840721 on Build GRCh38
rs949531043 Chr.20: 58840707-58840707 on Build GRCh38
rs868703522 Chr.20: 58840761-58840761 on Build GRCh38
rs751030899 Chr.20: 58840611-58840611 on Build GRCh38
rs753292441 Chr.20: 58840715-58840715 on Build GRCh38
rs745868121 Chr.20: 58840649-58840649 on Build GRCh38
rs75176432 Chr.20: 58840757-58840757 on Build GRCh38
rs78850964 Chr.20: 58840653-58840653 on Build GRCh38
rs748980591 Chr.20: 58840767-58840767 on Build GRCh38
rs772869359 Chr.20: 58840676-58840676 on Build GRCh38
rs373737048 Chr.20: 58840829-58840829 on Build GRCh38
rs76844956 Chr.20: 58840775-58840775 on Build GRCh38
rs768082058 Chr.20: 58840612-58840612 on Build GRCh38
rs746049514 Chr.20: 58840624-58840625 on Build GRCh38
rs781477248 Chr.20: 58840635-58840635 on Build GRCh38
rs953102685 Chr.20: 58840622-58840622 on Build GRCh38
rs769628289 Chr.20: 58840667-58840667 on Build GRCh38
rs764628148 Chr.20: 58840819-58840819 on Build GRCh38
rs79527543 Chr.20: 58840821-58840821 on Build GRCh38
rs747919997 Chr.20: 58840737-58840737 on Build GRCh38
rs376154152 Chr.20: 58840620-58840620 on Build GRCh38
rs770642877 Chr.20: 58840776-58840776 on Build GRCh38
rs867608721 Chr.20: 58840736-58840736 on Build GRCh38

View Full Product Details