Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 123010944-123011217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLVS2
Entrez Gene ID
134829
Gene Name
clavesin 2
Gene Aliases
C6orf212, C6orf213, RLBP1L2, bA160A10.4
UniGene
Hs.486361
Gene Chromosome Location
Chr.6: 122993693-123081282 on Build GRCh38
Transcripts
NM_001010852.3 XM_017010266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4403135 c.556G>A p.G186S Chr.6: 123011151-123011151 on Build GRCh38
54526 c.498C>T p.N166N Chr.6: 123011093-123011093 on Build GRCh38
739367 c.503C>A p.T168N Chr.6: 123011098-123011098 on Build GRCh38
7332682 c.487G>C p.D163H Chr.6: 123011082-123011082 on Build GRCh38
398601 c.415G>T p.A139S Chr.6: 123011010-123011010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909925415 Chr.6: 123011075-123011075 on Build GRCh38
rs778898775 Chr.6: 123011130-123011130 on Build GRCh38
rs745660729 Chr.6: 123010997-123010997 on Build GRCh38
rs745325386 Chr.6: 123010991-123010991 on Build GRCh38
rs771884601 Chr.6: 123011137-123011137 on Build GRCh38
rs770023200 Chr.6: 123011181-123011181 on Build GRCh38
rs375354563 Chr.6: 123011165-123011165 on Build GRCh38
rs745736656 Chr.6: 123011131-123011131 on Build GRCh38
rs766902762 Chr.6: 123011103-123011103 on Build GRCh38
rs776841053 Chr.6: 123011008-123011008 on Build GRCh38
rs374225032 Chr.6: 123010976-123010976 on Build GRCh38
rs759996346 Chr.6: 123011106-123011106 on Build GRCh38
rs368798011 Chr.6: 123010977-123010977 on Build GRCh38
rs773228597 Chr.6: 123011188-123011188 on Build GRCh38
rs770879812 Chr.6: 123010980-123010980 on Build GRCh38
rs984093628 Chr.6: 123010982-123010982 on Build GRCh38
rs980239020 Chr.6: 123011135-123011135 on Build GRCh38
rs771794699 Chr.6: 123010999-123010999 on Build GRCh38
rs779775195 Chr.6: 123011163-123011163 on Build GRCh38
rs761982510 Chr.6: 123011031-123011031 on Build GRCh38
rs765323473 Chr.6: 123011051-123011051 on Build GRCh38
rs755782858 Chr.6: 123011119-123011119 on Build GRCh38
rs781130103 Chr.6: 123010981-123010982 on Build GRCh38
rs763406760 Chr.6: 123011102-123011102 on Build GRCh38
rs920381124 Chr.6: 123011156-123011156 on Build GRCh38
rs147496902 Chr.6: 123011057-123011057 on Build GRCh38
rs201320099 Chr.6: 123011123-123011123 on Build GRCh38
rs200795338 Chr.6: 123011108-123011108 on Build GRCh38
rs371688821 Chr.6: 123011109-123011109 on Build GRCh38
rs751923428 Chr.6: 123011105-123011105 on Build GRCh38
rs139717598 Chr.6: 123011128-123011128 on Build GRCh38
rs773317056 Chr.6: 123011081-123011081 on Build GRCh38
rs376708439 Chr.6: 123011189-123011189 on Build GRCh38
rs144649146 Chr.6: 123010979-123010979 on Build GRCh38

View Full Product Details