Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102164639-102164876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R1
Entrez Gene ID
3554
Gene Name
interleukin 1 receptor type 1
Gene Aliases
CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80
UniGene
Hs.701982
Gene Chromosome Location
Chr.2: 102069638-102179874 on Build GRCh38
Transcripts
NM_000877.3 NM_001288706.1 NM_001320978.1 NM_001320980.1 NM_001320981.1 NM_001320982.1 NM_001320983.1 NM_001320984.1 NM_001320985.1 NM_001320986.1 XM_005263930.4 XM_005263934.4 XM_011511115.2 XM_011511116.1 XM_011511117.1 XM_011511118.2 XM_017003989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692236 c.124C>T p.R42C Chr.2: 102164836-102164836 on Build GRCh38
1579945 c.112G>A p.E38K Chr.2: 102164824-102164824 on Build GRCh38
1210542 c.125G>A p.R42H Chr.2: 102164837-102164837 on Build GRCh38
1579944 c.70A>C p.K24Q Chr.2: 102164782-102164782 on Build GRCh38
1176610 c.84A>C p.E28D Chr.2: 102164796-102164796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746323923 Chr.2: 102164831-102164831 on Build GRCh38
rs779664597 Chr.2: 102164789-102164789 on Build GRCh38
rs763351267 Chr.2: 102164841-102164841 on Build GRCh38
rs986186124 Chr.2: 102164705-102164705 on Build GRCh38
rs754771479 Chr.2: 102164795-102164795 on Build GRCh38
rs201587813 Chr.2: 102164816-102164816 on Build GRCh38
rs200886570 Chr.2: 102164764-102164764 on Build GRCh38
rs886956467 Chr.2: 102164693-102164693 on Build GRCh38
rs949809680 Chr.2: 102164679-102164679 on Build GRCh38
rs201457334 Chr.2: 102164733-102164733 on Build GRCh38
rs770360631 Chr.2: 102164834-102164834 on Build GRCh38
rs183879404 Chr.2: 102164686-102164686 on Build GRCh38
rs202008017 Chr.2: 102164775-102164775 on Build GRCh38
rs188550005 Chr.2: 102164840-102164840 on Build GRCh38
rs747874532 Chr.2: 102164818-102164818 on Build GRCh38
rs199520843 Chr.2: 102164756-102164756 on Build GRCh38
rs554579204 Chr.2: 102164704-102164704 on Build GRCh38
rs113620360 Chr.2: 102164793-102164793 on Build GRCh38
rs375255915 Chr.2: 102164747-102164747 on Build GRCh38
rs867221636 Chr.2: 102164836-102164836 on Build GRCh38
rs537771996 Chr.2: 102164745-102164745 on Build GRCh38
rs772861436 Chr.2: 102164825-102164825 on Build GRCh38
rs750296992 Chr.2: 102164783-102164783 on Build GRCh38
rs771640827 Chr.2: 102164821-102164821 on Build GRCh38
rs3917285 Chr.2: 102164706-102164706 on Build GRCh38
rs56337419 Chr.2: 102164788-102164788 on Build GRCh38

View Full Product Details