Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40698535-40698770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT24
Entrez Gene ID
192666
Gene Name
keratin 24
Gene Aliases
K24, KA24
UniGene
Hs.87383
Gene Chromosome Location
Chr.17: 40694246-40703750 on Build GRCh38
Transcripts
NM_019016.2 XM_006721739.3 XM_011524460.2 XM_017024299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383004 c.1398A>G p.S466S Chr.17: 40698614-40698614 on Build GRCh38
3517076 c.1422G>A p.R474R Chr.17: 40698590-40698590 on Build GRCh38
560895 c.1413G>T p.M471I Chr.17: 40698599-40698599 on Build GRCh38
138099 c.1472G>A p.R491Q Chr.17: 40698540-40698540 on Build GRCh38
5722609 c.1469G>A p.G490E Chr.17: 40698543-40698543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755771642 Chr.17: 40698677-40698677 on Build GRCh38
rs775327118 Chr.17: 40698570-40698570 on Build GRCh38
rs374325529 Chr.17: 40698696-40698696 on Build GRCh38
rs747547032 Chr.17: 40698558-40698558 on Build GRCh38
rs186682225 Chr.17: 40698709-40698709 on Build GRCh38
rs35992651 Chr.17: 40698652-40698653 on Build GRCh38
rs752005669 Chr.17: 40698627-40698627 on Build GRCh38
rs748971280 Chr.17: 40698694-40698694 on Build GRCh38
rs202143442 Chr.17: 40698565-40698565 on Build GRCh38
rs866964813 Chr.17: 40698590-40698590 on Build GRCh38
rs966287926 Chr.17: 40698703-40698703 on Build GRCh38
rs761851135 Chr.17: 40698602-40698602 on Build GRCh38
rs1015881129 Chr.17: 40698722-40698722 on Build GRCh38
rs762604288 Chr.17: 40698577-40698577 on Build GRCh38
rs984851595 Chr.17: 40698727-40698727 on Build GRCh38
rs373036343 Chr.17: 40698616-40698616 on Build GRCh38
rs752988516 Chr.17: 40698621-40698621 on Build GRCh38
rs780693669 Chr.17: 40698564-40698564 on Build GRCh38
rs143897454 Chr.17: 40698626-40698626 on Build GRCh38
rs753724177 Chr.17: 40698715-40698715 on Build GRCh38
rs757823304 Chr.17: 40698649-40698649 on Build GRCh38
rs764051380 Chr.17: 40698683-40698683 on Build GRCh38
rs757106465 Chr.17: 40698720-40698720 on Build GRCh38
rs756027875 Chr.17: 40698604-40698604 on Build GRCh38
rs754038118 Chr.17: 40698612-40698612 on Build GRCh38
rs770361757 Chr.17: 40698559-40698559 on Build GRCh38
rs774360695 Chr.17: 40698589-40698589 on Build GRCh38
rs183907017 Chr.17: 40698670-40698670 on Build GRCh38
rs769403420 Chr.17: 40698566-40698566 on Build GRCh38
rs750702894 Chr.17: 40698714-40698714 on Build GRCh38
rs952706400 Chr.17: 40698710-40698710 on Build GRCh38
rs768524198 Chr.17: 40698583-40698583 on Build GRCh38
rs142345813 Chr.17: 40698619-40698619 on Build GRCh38
rs766553742 Chr.17: 40698603-40698603 on Build GRCh38

View Full Product Details