Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83971813-83972086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPK
Entrez Gene ID
3190
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
AUKS, CSBP, HNRPK, TUNP
UniGene
Hs.522257
Gene Chromosome Location
Chr.9: 83968083-83980782 on Build GRCh38
Transcripts
NM_001318186.1 NM_001318187.1 NM_001318188.1 NM_002140.4 NM_031262.3 NM_031263.3 XM_005251960.2 XM_005251963.3 XM_005251965.2 XM_011518616.1 XM_017014668.1 XM_017014669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7151227 c.899_900insG p.G301fs*2 Chr.9: 83971935-83971936 on Build GRCh38
3659223 c.706C>T p.R236W Chr.9: 83971922-83971922 on Build GRCh38
3659226 c.821C>T p.P274L Chr.9: 83972014-83972014 on Build GRCh38
4785081 c.933A>T p.P311P Chr.9: 83971902-83971902 on Build GRCh38
4747588 c.844_846delGAT p.D282delD Chr.9: 83971989-83971991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757092061 Chr.9: 83972056-83972056 on Build GRCh38
rs771433348 Chr.9: 83971937-83971937 on Build GRCh38
rs765696098 Chr.9: 83971891-83971893 on Build GRCh38
rs760799303 Chr.9: 83971853-83971854 on Build GRCh38
rs967239711 Chr.9: 83971953-83971953 on Build GRCh38
rs754372494 Chr.9: 83971848-83971848 on Build GRCh38
rs765773838 Chr.9: 83971962-83971962 on Build GRCh38
rs199832698 Chr.9: 83971868-83971868 on Build GRCh38
rs267602294 Chr.9: 83972034-83972034 on Build GRCh38
rs552959343 Chr.9: 83971861-83971861 on Build GRCh38
rs766331617 Chr.9: 83971994-83971994 on Build GRCh38
rs762252496 Chr.9: 83971849-83971849 on Build GRCh38
rs375311945 Chr.9: 83971957-83971957 on Build GRCh38
rs755210140 Chr.9: 83972024-83972024 on Build GRCh38
rs960183341 Chr.9: 83972043-83972043 on Build GRCh38
rs370764430 Chr.9: 83971859-83971859 on Build GRCh38
rs745419757 Chr.9: 83971924-83971924 on Build GRCh38
rs771055929 Chr.9: 83971885-83971885 on Build GRCh38
rs898038676 Chr.9: 83971842-83971842 on Build GRCh38
rs572386998 Chr.9: 83971865-83971865 on Build GRCh38
rs866186478 Chr.9: 83971971-83971971 on Build GRCh38
rs544665964 Chr.9: 83971865-83971867 on Build GRCh38
rs769080208 Chr.9: 83971950-83971950 on Build GRCh38
rs753722223 Chr.9: 83972039-83972039 on Build GRCh38
rs764324655 Chr.9: 83971846-83971846 on Build GRCh38
rs879255263 Chr.9: 83971903-83971904 on Build GRCh38
rs267602293 Chr.9: 83971963-83971963 on Build GRCh38
rs751813868 Chr.9: 83972016-83972016 on Build GRCh38
rs981401210 Chr.9: 83972040-83972040 on Build GRCh38
rs546137739 Chr.9: 83971939-83971939 on Build GRCh38
rs777039431 Chr.9: 83971952-83971952 on Build GRCh38
rs544151579 Chr.9: 83971865-83971866 on Build GRCh38
rs766661308 Chr.9: 83971856-83971857 on Build GRCh38
rs758707867 Chr.9: 83971989-83971991 on Build GRCh38
rs761122385 Chr.9: 83971843-83971843 on Build GRCh38
rs1008046487 Chr.9: 83971884-83971884 on Build GRCh38
rs749075045 Chr.9: 83971876-83971876 on Build GRCh38
rs763046990 Chr.9: 83971992-83971992 on Build GRCh38
rs557886047 Chr.9: 83972068-83972068 on Build GRCh38
rs1032464428 Chr.9: 83971894-83971894 on Build GRCh38
rs777367009 Chr.9: 83971871-83971871 on Build GRCh38
rs778958822 Chr.9: 83971905-83971905 on Build GRCh38
rs863223402 Chr.9: 83971880-83971881 on Build GRCh38
rs750911102 Chr.9: 83971987-83971987 on Build GRCh38
rs755972219 Chr.9: 83971870-83971870 on Build GRCh38
rs760176912 Chr.9: 83971943-83971943 on Build GRCh38
rs781469290 Chr.9: 83972025-83972025 on Build GRCh38
rs886041807 Chr.9: 83972055-83972056 on Build GRCh38
rs761993593 Chr.9: 83971853-83971853 on Build GRCh38
rs756454668 Chr.9: 83971854-83971856 on Build GRCh38
rs753210017 Chr.9: 83971954-83971956 on Build GRCh38

View Full Product Details