Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70251712-70251961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC6
Entrez Gene ID
118460
Gene Name
exosome component 6
Gene Aliases
EAP4, MTR3, Mtr3p, hMtr3p, p11
UniGene
Hs.660633
Gene Chromosome Location
Chr.16: 70250231-70251930 on Build GRCh38
Transcripts
NM_058219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3511341 c.9G>A p.G3G Chr.16: 70251892-70251892 on Build GRCh38
4682074 c.72G>C p.E24D Chr.16: 70251829-70251829 on Build GRCh38
5971893 c.114C>T p.P38P Chr.16: 70251787-70251787 on Build GRCh38
4609705 c.143_168del26 p.Q48fs*65 Chr.16: 70251733-70251758 on Build GRCh38
4987364 c.140G>A p.S47N Chr.16: 70251761-70251761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368250236 Chr.16: 70251853-70251853 on Build GRCh38
rs753040972 Chr.16: 70251891-70251891 on Build GRCh38
rs776157604 Chr.16: 70251822-70251822 on Build GRCh38
rs1031268602 Chr.16: 70251756-70251756 on Build GRCh38
rs775215603 Chr.16: 70251872-70251872 on Build GRCh38
rs746467249 Chr.16: 70251859-70251859 on Build GRCh38
rs779482874 Chr.16: 70251805-70251805 on Build GRCh38
rs774270135 Chr.16: 70251911-70251911 on Build GRCh38
rs180927891 Chr.16: 70251914-70251914 on Build GRCh38
rs570335967 Chr.16: 70251923-70251923 on Build GRCh38
rs956832401 Chr.16: 70251772-70251772 on Build GRCh38
rs562015262 Chr.16: 70251879-70251879 on Build GRCh38
rs968961091 Chr.16: 70251748-70251748 on Build GRCh38
rs757822691 Chr.16: 70251902-70251902 on Build GRCh38
rs748664208 Chr.16: 70251811-70251811 on Build GRCh38
rs765581279 Chr.16: 70251885-70251885 on Build GRCh38
rs747461947 Chr.16: 70251899-70251899 on Build GRCh38
rs768916853 Chr.16: 70251803-70251803 on Build GRCh38
rs952685621 Chr.16: 70251929-70251929 on Build GRCh38
rs776528205 Chr.16: 70251918-70251918 on Build GRCh38
rs770546892 Chr.16: 70251821-70251821 on Build GRCh38
rs934330699 Chr.16: 70251898-70251898 on Build GRCh38
rs753994231 Chr.16: 70251882-70251882 on Build GRCh38
rs527466660 Chr.16: 70251908-70251908 on Build GRCh38
rs759251046 Chr.16: 70251761-70251761 on Build GRCh38
rs769394451 Chr.16: 70251904-70251904 on Build GRCh38
rs201044031 Chr.16: 70251812-70251812 on Build GRCh38
rs781217289 Chr.16: 70251791-70251791 on Build GRCh38
rs924983120 Chr.16: 70251793-70251793 on Build GRCh38
rs761629603 Chr.16: 70251912-70251912 on Build GRCh38
rs566681785 Chr.16: 70251903-70251903 on Build GRCh38
rs564587248 Chr.16: 70251873-70251873 on Build GRCh38
rs776267407 Chr.16: 70251869-70251869 on Build GRCh38
rs572594008 Chr.16: 70251788-70251788 on Build GRCh38
rs753625579 Chr.16: 70251760-70251760 on Build GRCh38
rs750476872 Chr.16: 70251840-70251840 on Build GRCh38
rs776639358 Chr.16: 70251871-70251871 on Build GRCh38
rs1013769970 Chr.16: 70251785-70251785 on Build GRCh38
rs1025173889 Chr.16: 70251913-70251913 on Build GRCh38
rs554205982 Chr.16: 70251784-70251784 on Build GRCh38
rs758757192 Chr.16: 70251934-70251934 on Build GRCh38
rs749026748 Chr.16: 70251909-70251909 on Build GRCh38
rs752951027 Chr.16: 70251926-70251926 on Build GRCh38
rs772828808 Chr.16: 70251817-70251817 on Build GRCh38
rs982770313 Chr.16: 70251932-70251932 on Build GRCh38
rs752527501 Chr.16: 70251835-70251835 on Build GRCh38
rs780172429 Chr.16: 70251800-70251800 on Build GRCh38
rs753992975 Chr.16: 70251847-70251847 on Build GRCh38
rs543954936 Chr.16: 70251877-70251877 on Build GRCh38
rs979067761 Chr.16: 70251769-70251769 on Build GRCh38
rs531804444 Chr.16: 70251876-70251876 on Build GRCh38
rs1027034329 Chr.16: 70251927-70251927 on Build GRCh38
rs759287255 Chr.16: 70251824-70251824 on Build GRCh38
rs147573939 Chr.16: 70251884-70251885 on Build GRCh38
rs761868365 Chr.16: 70251878-70251878 on Build GRCh38
rs758396305 Chr.16: 70251787-70251787 on Build GRCh38
rs919965574 Chr.16: 70251825-70251825 on Build GRCh38
rs758843378 Chr.16: 70251892-70251892 on Build GRCh38
rs756308308 Chr.16: 70251799-70251799 on Build GRCh38
rs748576023 Chr.16: 70251854-70251854 on Build GRCh38
rs760347512 Chr.16: 70251733-70251733 on Build GRCh38
rs796735241 Chr.16: 70251831-70251831 on Build GRCh38
rs745478628 Chr.16: 70251870-70251870 on Build GRCh38
rs762854436 Chr.16: 70251837-70251837 on Build GRCh38
rs778427187 Chr.16: 70251856-70251856 on Build GRCh38
rs769455021 Chr.16: 70251938-70251938 on Build GRCh38
rs1048304389 Chr.16: 70251927-70251928 on Build GRCh38
rs772661085 Chr.16: 70251855-70251855 on Build GRCh38
rs765999641 Chr.16: 70251745-70251745 on Build GRCh38
rs764330820 Chr.16: 70251942-70251942 on Build GRCh38
rs867552592 Chr.16: 70251839-70251839 on Build GRCh38
rs756153434 Chr.16: 70251844-70251844 on Build GRCh38
rs770329684 Chr.16: 70251862-70251862 on Build GRCh38
rs760131999 Chr.16: 70251820-70251820 on Build GRCh38
rs765495451 Chr.16: 70251925-70251925 on Build GRCh38
rs569157236 Chr.16: 70251868-70251868 on Build GRCh38
rs765012814 Chr.16: 70251832-70251832 on Build GRCh38
rs768427140 Chr.16: 70251910-70251910 on Build GRCh38
rs529346561 Chr.16: 70251887-70251887 on Build GRCh38
rs772927500 Chr.16: 70251729-70251729 on Build GRCh38
rs201962601 Chr.16: 70251896-70251896 on Build GRCh38
rs1048328129 Chr.16: 70251826-70251826 on Build GRCh38
rs982629731 Chr.16: 70251850-70251850 on Build GRCh38
rs755322940 Chr.16: 70251852-70251852 on Build GRCh38
rs976642772 Chr.16: 70251860-70251860 on Build GRCh38
rs765097066 Chr.16: 70251778-70251778 on Build GRCh38

View Full Product Details