Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 105490032-105490305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAOA
Entrez Gene ID
267012
Gene Name
D-amino acid oxidase activator
Gene Aliases
LG72, SG72
UniGene
Hs.381382
Gene Chromosome Location
Chr.13: 105465867-105491034 on Build GRCh38
Transcripts
NM_001161812.1 NM_001161814.1 NM_172370.4 XM_005254042.1 NR_040247.1

Gene Symbol
DAOA-AS1
Entrez Gene ID
282706
Gene Name
DAOA antisense RNA 1
Gene Aliases
DAOA-AS, DAOAAS, G30
UniGene
Hs.558713
Gene Chromosome Location
Chr.13: 105459055-105505681 on Build GRCh38
Transcripts
NM_001161812.1 NM_001161814.1 NM_172370.4 XM_005254042.1 NR_040247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945224 c.446C>A p.S149Y Chr.13: 105490065-105490065 on Build GRCh38
5897862 c.436G>A p.E146K Chr.13: 105490055-105490055 on Build GRCh38
280293 c.443C>A p.T148N Chr.13: 105490062-105490062 on Build GRCh38
166510 c.417delC p.Q140fs*8 Chr.13: 105490036-105490036 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376693801 Chr.13: 105490123-105490123 on Build GRCh38
rs760212075 Chr.13: 105490099-105490099 on Build GRCh38
rs184876865 Chr.13: 105490190-105490190 on Build GRCh38
rs927979572 Chr.13: 105490112-105490112 on Build GRCh38
rs561408705 Chr.13: 105490174-105490174 on Build GRCh38
rs938007449 Chr.13: 105490134-105490134 on Build GRCh38
rs370299059 Chr.13: 105490184-105490184 on Build GRCh38
rs764939690 Chr.13: 105490131-105490131 on Build GRCh38
rs995108905 Chr.13: 105490157-105490157 on Build GRCh38
rs563731892 Chr.13: 105490225-105490225 on Build GRCh38
rs905177666 Chr.13: 105490264-105490264 on Build GRCh38
rs763735781 Chr.13: 105490107-105490107 on Build GRCh38
rs776161548 Chr.13: 105490108-105490108 on Build GRCh38
rs367543081 Chr.13: 105490053-105490053 on Build GRCh38
rs147257124 Chr.13: 105490087-105490087 on Build GRCh38
rs375731437 Chr.13: 105490167-105490167 on Build GRCh38
rs1046428463 Chr.13: 105490111-105490111 on Build GRCh38
rs1055493148 Chr.13: 105490137-105490137 on Build GRCh38
rs995839679 Chr.13: 105490270-105490270 on Build GRCh38
rs1044737705 Chr.13: 105490246-105490246 on Build GRCh38
rs886248689 Chr.13: 105490189-105490189 on Build GRCh38
rs895657352 Chr.13: 105490228-105490228 on Build GRCh38
rs574977423 Chr.13: 105490186-105490186 on Build GRCh38
rs1018671738 Chr.13: 105490243-105490243 on Build GRCh38
rs964407087 Chr.13: 105490259-105490259 on Build GRCh38
rs72549494 Chr.13: 105490241-105490241 on Build GRCh38
rs899064738 Chr.13: 105490146-105490146 on Build GRCh38

View Full Product Details