Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55366159-55366432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LANCL2
Entrez Gene ID
55915
Gene Name
LanC like 2
Gene Aliases
GPR69B, TASP
UniGene
Hs.595384
Gene Chromosome Location
Chr.7: 55365448-55433742 on Build GRCh38
Transcripts
NM_018697.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285116 c.158C>A p.P53Q Chr.7: 55366183-55366183 on Build GRCh38
7329375 c.190C>T p.H64Y Chr.7: 55366215-55366215 on Build GRCh38
7287946 c.179G>A p.G60D Chr.7: 55366204-55366204 on Build GRCh38
6284628 c.184C>A p.P62T Chr.7: 55366209-55366209 on Build GRCh38
6699509 c.191A>G p.H64R Chr.7: 55366216-55366216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749936737 Chr.7: 55366269-55366269 on Build GRCh38
rs990623449 Chr.7: 55366312-55366312 on Build GRCh38
rs765895707 Chr.7: 55366307-55366307 on Build GRCh38
rs894904570 Chr.7: 55366372-55366372 on Build GRCh38
rs776374472 Chr.7: 55366203-55366203 on Build GRCh38
rs772898187 Chr.7: 55366255-55366255 on Build GRCh38
rs893694834 Chr.7: 55366339-55366339 on Build GRCh38
rs1053605934 Chr.7: 55366326-55366326 on Build GRCh38
rs1052027634 Chr.7: 55366197-55366197 on Build GRCh38
rs545468613 Chr.7: 55366349-55366349 on Build GRCh38
rs2272262 Chr.7: 55366187-55366187 on Build GRCh38
rs981386784 Chr.7: 55366276-55366276 on Build GRCh38
rs919300423 Chr.7: 55366275-55366275 on Build GRCh38
rs767726602 Chr.7: 55366267-55366267 on Build GRCh38
rs1022092783 Chr.7: 55366406-55366406 on Build GRCh38
rs1009513995 Chr.7: 55366341-55366341 on Build GRCh38
rs757966543 Chr.7: 55366282-55366282 on Build GRCh38
rs769399790 Chr.7: 55366253-55366253 on Build GRCh38
rs555675982 Chr.7: 55366245-55366245 on Build GRCh38
rs578040775 Chr.7: 55366305-55366305 on Build GRCh38
rs755478863 Chr.7: 55366375-55366375 on Build GRCh38
rs933346632 Chr.7: 55366389-55366389 on Build GRCh38
rs1020641109 Chr.7: 55366240-55366240 on Build GRCh38
rs954111855 Chr.7: 55366407-55366407 on Build GRCh38
rs757806770 Chr.7: 55366243-55366243 on Build GRCh38
rs549984075 Chr.7: 55366395-55366395 on Build GRCh38
rs375322160 Chr.7: 55366259-55366259 on Build GRCh38
rs912443831 Chr.7: 55366331-55366331 on Build GRCh38
rs774733352 Chr.7: 55366195-55366195 on Build GRCh38
rs373246574 Chr.7: 55366251-55366251 on Build GRCh38
rs748242301 Chr.7: 55366180-55366180 on Build GRCh38
rs910653072 Chr.7: 55366408-55366408 on Build GRCh38
rs1032103852 Chr.7: 55366289-55366289 on Build GRCh38
rs1003664424 Chr.7: 55366353-55366353 on Build GRCh38
rs1031949408 Chr.7: 55366398-55366398 on Build GRCh38
rs531498527 Chr.7: 55366379-55366379 on Build GRCh38
rs972383823 Chr.7: 55366383-55366383 on Build GRCh38
rs537364684 Chr.7: 55366215-55366215 on Build GRCh38
rs778209297 Chr.7: 55366236-55366236 on Build GRCh38
rs963958741 Chr.7: 55366377-55366377 on Build GRCh38
rs372067135 Chr.7: 55366182-55366182 on Build GRCh38
rs369008408 Chr.7: 55366223-55366223 on Build GRCh38
rs776142784 Chr.7: 55366200-55366200 on Build GRCh38
rs768301130 Chr.7: 55366249-55366249 on Build GRCh38
rs766254477 Chr.7: 55366216-55366216 on Build GRCh38
rs2272263 Chr.7: 55366191-55366191 on Build GRCh38
rs765219069 Chr.7: 55366208-55366208 on Build GRCh38
rs774127610 Chr.7: 55366261-55366261 on Build GRCh38
rs896800563 Chr.7: 55366352-55366352 on Build GRCh38
rs756167756 Chr.7: 55366225-55366225 on Build GRCh38
rs991639176 Chr.7: 55366368-55366368 on Build GRCh38
rs987485239 Chr.7: 55366404-55366404 on Build GRCh38
rs759261384 Chr.7: 55366264-55366264 on Build GRCh38
rs1003069184 Chr.7: 55366392-55366392 on Build GRCh38
rs762970382 Chr.7: 55366257-55366257 on Build GRCh38
rs764133170 Chr.7: 55366274-55366274 on Build GRCh38
rs956230412 Chr.7: 55366399-55366399 on Build GRCh38
rs564292519 Chr.7: 55366364-55366364 on Build GRCh38
rs933400318 Chr.7: 55366279-55366279 on Build GRCh38
rs1007394762 Chr.7: 55366205-55366205 on Build GRCh38
rs919214141 Chr.7: 55366385-55366385 on Build GRCh38
rs781439805 Chr.7: 55366376-55366376 on Build GRCh38
rs752762048 Chr.7: 55366224-55366224 on Build GRCh38
rs190130774 Chr.7: 55366196-55366196 on Build GRCh38
rs768263181 Chr.7: 55366199-55366199 on Build GRCh38
rs778264159 Chr.7: 55366183-55366183 on Build GRCh38
rs559689533 Chr.7: 55366302-55366302 on Build GRCh38
rs541111797 Chr.7: 55366252-55366252 on Build GRCh38
rs985297060 Chr.7: 55366327-55366327 on Build GRCh38
rs756364478 Chr.7: 55366271-55366271 on Build GRCh38
rs781318029 Chr.7: 55366179-55366179 on Build GRCh38
rs749761080 Chr.7: 55366238-55366238 on Build GRCh38
rs562011202 Chr.7: 55366413-55366413 on Build GRCh38
rs1033442727 Chr.7: 55366355-55366355 on Build GRCh38
rs779249861 Chr.7: 55366244-55366244 on Build GRCh38
rs1044562804 Chr.7: 55366381-55366381 on Build GRCh38
rs374837911 Chr.7: 55366220-55366220 on Build GRCh38
rs957670474 Chr.7: 55366217-55366217 on Build GRCh38
rs970457801 Chr.7: 55366350-55366350 on Build GRCh38
rs1032225986 Chr.7: 55366210-55366210 on Build GRCh38

View Full Product Details