Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62860472-62860717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP1
Entrez Gene ID
23239
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
PHLPP, PLEKHE1, PPM3A, SCOP
UniGene
Hs.465337
Gene Chromosome Location
Chr.18: 62715439-62980443 on Build GRCh38
Transcripts
NM_194449.3 XM_011525886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573698 c.396_400delGTGTT p.C133fs*1 Chr.18: 62860470-62860474 on Build GRCh38
1189720 c.432C>A p.F144L Chr.18: 62860506-62860506 on Build GRCh38
6557098 c.1948G>T p.E650* Chr.18: 62860483-62860483 on Build GRCh38
6745542 c.1981G>T p.D661Y Chr.18: 62860516-62860516 on Build GRCh38
4789500 c.413A>G p.H138R Chr.18: 62860487-62860487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374899951 Chr.18: 62860583-62860583 on Build GRCh38
rs776692723 Chr.18: 62860590-62860590 on Build GRCh38
rs780009916 Chr.18: 62860622-62860622 on Build GRCh38
rs756273477 Chr.18: 62860617-62860617 on Build GRCh38
rs745989631 Chr.18: 62860515-62860515 on Build GRCh38
rs771735382 Chr.18: 62860572-62860572 on Build GRCh38
rs752558485 Chr.18: 62860601-62860601 on Build GRCh38
rs187962936 Chr.18: 62860566-62860566 on Build GRCh38
rs772829773 Chr.18: 62860576-62860576 on Build GRCh38
rs879074888 Chr.18: 62860643-62860643 on Build GRCh38
rs756229770 Chr.18: 62860523-62860523 on Build GRCh38
rs367681267 Chr.18: 62860607-62860607 on Build GRCh38
rs754872613 Chr.18: 62860506-62860506 on Build GRCh38
rs760220479 Chr.18: 62860582-62860582 on Build GRCh38
rs957408746 Chr.18: 62860646-62860646 on Build GRCh38
rs764931071 Chr.18: 62860508-62860508 on Build GRCh38
rs762816249 Chr.18: 62860604-62860604 on Build GRCh38
rs781511873 Chr.18: 62860514-62860514 on Build GRCh38
rs767106617 Chr.18: 62860662-62860662 on Build GRCh38
rs755471039 Chr.18: 62860632-62860632 on Build GRCh38
rs780674753 Chr.18: 62860544-62860544 on Build GRCh38
rs769147523 Chr.18: 62860561-62860561 on Build GRCh38
rs183375168 Chr.18: 62860510-62860510 on Build GRCh38
rs894285699 Chr.18: 62860654-62860656 on Build GRCh38
rs192314431 Chr.18: 62860610-62860610 on Build GRCh38
rs749743970 Chr.18: 62860556-62860556 on Build GRCh38
rs761878920 Chr.18: 62860679-62860679 on Build GRCh38
rs757561972 Chr.18: 62860512-62860512 on Build GRCh38
rs569299328 Chr.18: 62860499-62860499 on Build GRCh38
rs1011377337 Chr.18: 62860657-62860657 on Build GRCh38
rs765159319 Chr.18: 62860598-62860598 on Build GRCh38
rs766389894 Chr.18: 62860495-62860495 on Build GRCh38
rs779428105 Chr.18: 62860638-62860638 on Build GRCh38
rs572064997 Chr.18: 62860596-62860596 on Build GRCh38
rs375891044 Chr.18: 62860624-62860624 on Build GRCh38

View Full Product Details