Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64317290-64317551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT112
Entrez Gene ID
51504
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Gene Aliases
HSPC152, HSPC170, TRM112, TRMT11-2, hTrm112
UniGene
Hs.333579
Gene Chromosome Location
Chr.11: 64316492-64318084 on Build GRCh38
Transcripts
NM_001286082.1 NM_001286084.1 NM_016404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5990999 c.17A>G p.H6R Chr.11: 64317510-64317510 on Build GRCh38
3719457 c.42G>A p.R14R Chr.11: 64317485-64317485 on Build GRCh38
5033564 c.78+3A>C p.? Chr.11: 64317446-64317446 on Build GRCh38
1628146 c.20A>G p.N7S Chr.11: 64317507-64317507 on Build GRCh38
4035109 c.117C>T p.P39P Chr.11: 64317327-64317327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375491426 Chr.11: 64317413-64317413 on Build GRCh38
rs768451942 Chr.11: 64317359-64317359 on Build GRCh38
rs777383647 Chr.11: 64317440-64317440 on Build GRCh38
rs550258200 Chr.11: 64317368-64317368 on Build GRCh38
rs530548412 Chr.11: 64317328-64317328 on Build GRCh38
rs769642779 Chr.11: 64317480-64317480 on Build GRCh38
rs981855947 Chr.11: 64317411-64317411 on Build GRCh38
rs749039296 Chr.11: 64317507-64317507 on Build GRCh38
rs775252346 Chr.11: 64317482-64317482 on Build GRCh38
rs1044184981 Chr.11: 64317511-64317511 on Build GRCh38
rs539630674 Chr.11: 64317387-64317387 on Build GRCh38
rs775436432 Chr.11: 64317460-64317460 on Build GRCh38
rs377669870 Chr.11: 64317506-64317506 on Build GRCh38
rs763156167 Chr.11: 64317403-64317403 on Build GRCh38
rs780565402 Chr.11: 64317421-64317421 on Build GRCh38
rs1056063179 Chr.11: 64317475-64317475 on Build GRCh38
rs757689139 Chr.11: 64317347-64317347 on Build GRCh38
rs747870648 Chr.11: 64317436-64317436 on Build GRCh38
rs968285613 Chr.11: 64317430-64317430 on Build GRCh38
rs769802254 Chr.11: 64317381-64317387 on Build GRCh38
rs745863361 Chr.11: 64317499-64317499 on Build GRCh38
rs994009920 Chr.11: 64317532-64317532 on Build GRCh38
rs200904021 Chr.11: 64317487-64317487 on Build GRCh38
rs776522864 Chr.11: 64317398-64317398 on Build GRCh38
rs751566570 Chr.11: 64317409-64317409 on Build GRCh38
rs750834611 Chr.11: 64317416-64317416 on Build GRCh38
rs998108042 Chr.11: 64317367-64317367 on Build GRCh38
rs986455749 Chr.11: 64317336-64317336 on Build GRCh38
rs552847770 Chr.11: 64317401-64317401 on Build GRCh38
rs774517266 Chr.11: 64317483-64317483 on Build GRCh38
rs28699214 Chr.11: 64317412-64317412 on Build GRCh38
rs535716108 Chr.11: 64317486-64317486 on Build GRCh38
rs370461838 Chr.11: 64317512-64317512 on Build GRCh38
rs768943059 Chr.11: 64317508-64317508 on Build GRCh38
rs771779655 Chr.11: 64317369-64317369 on Build GRCh38
rs757820595 Chr.11: 64317410-64317410 on Build GRCh38
rs941163341 Chr.11: 64317514-64317514 on Build GRCh38
rs770924814 Chr.11: 64317462-64317462 on Build GRCh38
rs759280010 Chr.11: 64317400-64317400 on Build GRCh38
rs762738556 Chr.11: 64317480-64317481 on Build GRCh38
rs764289286 Chr.11: 64317405-64317405 on Build GRCh38
rs758101903 Chr.11: 64317439-64317439 on Build GRCh38
rs759517900 Chr.11: 64317316-64317316 on Build GRCh38
rs140189524 Chr.11: 64317384-64317384 on Build GRCh38
rs562975998 Chr.11: 64317530-64317530 on Build GRCh38
rs746464116 Chr.11: 64317445-64317445 on Build GRCh38
rs756469761 Chr.11: 64317515-64317515 on Build GRCh38
rs756653486 Chr.11: 64317491-64317491 on Build GRCh38
rs754279340 Chr.11: 64317521-64317521 on Build GRCh38
rs138907479 Chr.11: 64317460-64317460 on Build GRCh38
rs745665266 Chr.11: 64317479-64317479 on Build GRCh38
rs574861213 Chr.11: 64317497-64317497 on Build GRCh38
rs41294430 Chr.11: 64317534-64317534 on Build GRCh38
rs761305189 Chr.11: 64317517-64317517 on Build GRCh38
rs766953448 Chr.11: 64317518-64317518 on Build GRCh38
rs770744162 Chr.11: 64317392-64317392 on Build GRCh38
rs200361095 Chr.11: 64317526-64317526 on Build GRCh38
rs1030379288 Chr.11: 64317533-64317533 on Build GRCh38
rs781653814 Chr.11: 64317353-64317353 on Build GRCh38
rs780435157 Chr.11: 64317510-64317510 on Build GRCh38
rs566781863 Chr.11: 64317419-64317419 on Build GRCh38
rs776373517 Chr.11: 64317471-64317471 on Build GRCh38
rs146953593 Chr.11: 64317337-64317337 on Build GRCh38
rs759763606 Chr.11: 64317522-64317522 on Build GRCh38
rs751041580 Chr.11: 64317484-64317484 on Build GRCh38
rs910462138 Chr.11: 64317446-64317446 on Build GRCh38
rs763716858 Chr.11: 64317481-64317481 on Build GRCh38
rs778256368 Chr.11: 64317316-64317316 on Build GRCh38
rs570596497 Chr.11: 64317378-64317378 on Build GRCh38
rs778654155 Chr.11: 64317365-64317365 on Build GRCh38
rs913551366 Chr.11: 64317394-64317394 on Build GRCh38
rs375653671 Chr.11: 64317485-64317485 on Build GRCh38
rs938822025 Chr.11: 64317461-64317461 on Build GRCh38
rs746184459 Chr.11: 64317354-64317354 on Build GRCh38
rs750819853 Chr.11: 64317379-64317379 on Build GRCh38
rs1037280445 Chr.11: 64317515-64317516 on Build GRCh38

View Full Product Details