Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 224924-225185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THOC1
Entrez Gene ID
9984
Gene Name
THO complex 1
Gene Aliases
HPR1, P84, P84N5
UniGene
Hs.712543
Gene Chromosome Location
Chr.18: 214348-268059 on Build GRCh38
Transcripts
NM_005131.2 XM_011525772.2 XM_011525773.1 XM_011525774.2 XM_017026103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7393755 c.1169C>T p.S390L Chr.18: 224963-224963 on Build GRCh38
5473991 c.1188C>T p.C396C Chr.18: 224944-224944 on Build GRCh38
6577876 c.1208G>T p.R403I Chr.18: 224924-224924 on Build GRCh38
4444635 c.1162T>C p.W388R Chr.18: 224970-224970 on Build GRCh38
6016916 c.1125A>G p.S375S Chr.18: 225101-225101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7227515 Chr.18: 225051-225051 on Build GRCh38
rs563539953 Chr.18: 224962-224962 on Build GRCh38
rs374193233 Chr.18: 225101-225101 on Build GRCh38
rs749332430 Chr.18: 225118-225118 on Build GRCh38
rs772631557 Chr.18: 225055-225055 on Build GRCh38
rs183259365 Chr.18: 225116-225116 on Build GRCh38
rs201689148 Chr.18: 224984-224984 on Build GRCh38
rs929042081 Chr.18: 225085-225085 on Build GRCh38
rs140489878 Chr.18: 225027-225027 on Build GRCh38
rs543467899 Chr.18: 225017-225017 on Build GRCh38
rs915835551 Chr.18: 225045-225045 on Build GRCh38
rs769182418 Chr.18: 224982-224982 on Build GRCh38
rs71352928 Chr.18: 225081-225081 on Build GRCh38
rs779457153 Chr.18: 225134-225134 on Build GRCh38
rs546555244 Chr.18: 225052-225052 on Build GRCh38
rs776285687 Chr.18: 225015-225015 on Build GRCh38
rs759264119 Chr.18: 225023-225023 on Build GRCh38
rs530965977 Chr.18: 225005-225005 on Build GRCh38
rs779295121 Chr.18: 225011-225011 on Build GRCh38
rs778021280 Chr.18: 225118-225119 on Build GRCh38
rs865911913 Chr.18: 224979-224979 on Build GRCh38
rs756362305 Chr.18: 225108-225108 on Build GRCh38
rs368677132 Chr.18: 225119-225119 on Build GRCh38
rs376053242 Chr.18: 224963-224963 on Build GRCh38
rs748616386 Chr.18: 225149-225149 on Build GRCh38
rs751521866 Chr.18: 225070-225070 on Build GRCh38
rs111499826 Chr.18: 225010-225010 on Build GRCh38
rs780864058 Chr.18: 225086-225086 on Build GRCh38
rs762233590 Chr.18: 225009-225009 on Build GRCh38
rs756443918 Chr.18: 225006-225009 on Build GRCh38

View Full Product Details