Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4448176-4448421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPNS3
Entrez Gene ID
201305
Gene Name
sphingolipid transporter 3 (putative)
Gene Aliases
-
UniGene
Hs.657543
Gene Chromosome Location
Chr.17: 4433924-4488204 on Build GRCh38
Transcripts
NM_001320449.1 NM_182538.4 XM_011523723.1 XM_011523724.1 XM_011523725.1 XM_011523726.2 XM_011523728.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686735 c.770+1G>T p.? Chr.17: 4448304-4448304 on Build GRCh38
5586676 c.696C>T p.A232A Chr.17: 4448229-4448229 on Build GRCh38
2695274 c.731G>A p.R244K Chr.17: 4448264-4448264 on Build GRCh38
4067101 c.766A>G p.K256E Chr.17: 4448299-4448299 on Build GRCh38
4613452 c.766delA p.N257fs*11 Chr.17: 4448299-4448299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371109204 Chr.17: 4448237-4448237 on Build GRCh38
rs770809118 Chr.17: 4448263-4448263 on Build GRCh38
rs562273062 Chr.17: 4448251-4448251 on Build GRCh38
rs369580957 Chr.17: 4448284-4448284 on Build GRCh38
rs553060973 Chr.17: 4448399-4448399 on Build GRCh38
rs762439491 Chr.17: 4448302-4448302 on Build GRCh38
rs528953104 Chr.17: 4448364-4448364 on Build GRCh38
rs976767291 Chr.17: 4448365-4448365 on Build GRCh38
rs922703282 Chr.17: 4448370-4448370 on Build GRCh38
rs993157765 Chr.17: 4448274-4448274 on Build GRCh38
rs567810323 Chr.17: 4448320-4448320 on Build GRCh38
rs145332257 Chr.17: 4448208-4448208 on Build GRCh38
rs267604923 Chr.17: 4448228-4448228 on Build GRCh38
rs758468796 Chr.17: 4448321-4448321 on Build GRCh38
rs376371811 Chr.17: 4448283-4448283 on Build GRCh38
rs760341690 Chr.17: 4448290-4448290 on Build GRCh38
rs534788954 Chr.17: 4448392-4448392 on Build GRCh38
rs777568288 Chr.17: 4448338-4448338 on Build GRCh38
rs200186002 Chr.17: 4448215-4448215 on Build GRCh38
rs12450838 Chr.17: 4448265-4448265 on Build GRCh38
rs200134393 Chr.17: 4448336-4448336 on Build GRCh38
rs139104578 Chr.17: 4448242-4448242 on Build GRCh38
rs764601606 Chr.17: 4448293-4448293 on Build GRCh38
rs908487309 Chr.17: 4448330-4448330 on Build GRCh38
rs775040538 Chr.17: 4448354-4448354 on Build GRCh38
rs765768389 Chr.17: 4448310-4448310 on Build GRCh38
rs766479806 Chr.17: 4448322-4448322 on Build GRCh38
rs368376768 Chr.17: 4448352-4448352 on Build GRCh38
rs149895416 Chr.17: 4448248-4448248 on Build GRCh38
rs753111287 Chr.17: 4448216-4448216 on Build GRCh38
rs749043512 Chr.17: 4448343-4448343 on Build GRCh38
rs901247903 Chr.17: 4448230-4448230 on Build GRCh38
rs61740736 Chr.17: 4448239-4448239 on Build GRCh38
rs775071687 Chr.17: 4448289-4448289 on Build GRCh38
rs754434764 Chr.17: 4448227-4448227 on Build GRCh38
rs764514399 Chr.17: 4448219-4448219 on Build GRCh38
rs777515199 Chr.17: 4448250-4448250 on Build GRCh38
rs181393572 Chr.17: 4448344-4448344 on Build GRCh38
rs541963025 Chr.17: 4448244-4448244 on Build GRCh38
rs750540559 Chr.17: 4448313-4448313 on Build GRCh38
rs201210465 Chr.17: 4448300-4448300 on Build GRCh38
rs141783753 Chr.17: 4448229-4448229 on Build GRCh38
rs76531347 Chr.17: 4448377-4448377 on Build GRCh38
rs884250 Chr.17: 4448345-4448345 on Build GRCh38
rs745813090 Chr.17: 4448346-4448346 on Build GRCh38
rs565815972 Chr.17: 4448380-4448380 on Build GRCh38
rs533485889 Chr.17: 4448342-4448342 on Build GRCh38
rs763206458 Chr.17: 4448332-4448333 on Build GRCh38
rs200982219 Chr.17: 4448218-4448218 on Build GRCh38
rs751771660 Chr.17: 4448327-4448327 on Build GRCh38

View Full Product Details