Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43665249-43665522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAUR
Entrez Gene ID
5329
Gene Name
plasminogen activator, urokinase receptor
Gene Aliases
CD87, U-PAR, UPAR, URKR
UniGene
Hs.466871
Gene Chromosome Location
Chr.19: 43646095-43670346 on Build GRCh38
Transcripts
NM_001005376.2 NM_001005377.2 NM_001301037.1 NM_002659.3 XM_005258990.4 XM_011527027.2 XM_011527028.2 XM_011527029.1 XM_011527030.2 XM_011527031.2 XM_017026872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
389303 c.190G>T p.E64* Chr.19: 43665436-43665436 on Build GRCh38
712524 c.209C>G p.S70* Chr.19: 43665417-43665417 on Build GRCh38
6994867 c.231G>T p.L77L Chr.19: 43665395-43665395 on Build GRCh38
712525 c.209C>G p.S70* Chr.19: 43665417-43665417 on Build GRCh38
1481171 c.198C>G p.S66R Chr.19: 43665428-43665428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761858444 Chr.19: 43665278-43665278 on Build GRCh38
rs756019389 Chr.19: 43665302-43665302 on Build GRCh38
rs745738157 Chr.19: 43665371-43665371 on Build GRCh38
rs4251832 Chr.19: 43665313-43665313 on Build GRCh38
rs777730913 Chr.19: 43665409-43665409 on Build GRCh38
rs766700898 Chr.19: 43665471-43665471 on Build GRCh38
rs748557493 Chr.19: 43665448-43665448 on Build GRCh38
rs771741460 Chr.19: 43665325-43665325 on Build GRCh38
rs746689316 Chr.19: 43665502-43665502 on Build GRCh38
rs1019829785 Chr.19: 43665462-43665462 on Build GRCh38
rs774473988 Chr.19: 43665472-43665472 on Build GRCh38
rs781088559 Chr.19: 43665289-43665289 on Build GRCh38
rs752572976 Chr.19: 43665290-43665290 on Build GRCh38
rs779650701 Chr.19: 43665500-43665500 on Build GRCh38
rs146937073 Chr.19: 43665450-43665450 on Build GRCh38
rs377171020 Chr.19: 43665280-43665280 on Build GRCh38
rs372236539 Chr.19: 43665475-43665475 on Build GRCh38
rs184935465 Chr.19: 43665358-43665358 on Build GRCh38
rs11546138 Chr.19: 43665404-43665404 on Build GRCh38
rs530787123 Chr.19: 43665422-43665422 on Build GRCh38
rs371000505 Chr.19: 43665449-43665449 on Build GRCh38
rs1020749878 Chr.19: 43665438-43665438 on Build GRCh38
rs753884538 Chr.19: 43665408-43665408 on Build GRCh38
rs749924157 Chr.19: 43665498-43665498 on Build GRCh38
rs546374331 Chr.19: 43665355-43665355 on Build GRCh38
rs146202545 Chr.19: 43665359-43665359 on Build GRCh38
rs144494060 Chr.19: 43665365-43665365 on Build GRCh38
rs371081407 Chr.19: 43665476-43665476 on Build GRCh38
rs777177825 Chr.19: 43665443-43665443 on Build GRCh38
rs200062790 Chr.19: 43665483-43665483 on Build GRCh38
rs977635101 Chr.19: 43665387-43665387 on Build GRCh38
rs753768463 Chr.19: 43665420-43665420 on Build GRCh38
rs771657400 Chr.19: 43665429-43665429 on Build GRCh38
rs557064860 Chr.19: 43665285-43665285 on Build GRCh38
rs562436214 Chr.19: 43665377-43665377 on Build GRCh38
rs779740725 Chr.19: 43665430-43665430 on Build GRCh38
rs778513898 Chr.19: 43665319-43665319 on Build GRCh38
rs763230958 Chr.19: 43665369-43665369 on Build GRCh38
rs757972378 Chr.19: 43665499-43665499 on Build GRCh38
rs747395993 Chr.19: 43665431-43665431 on Build GRCh38
rs371882099 Chr.19: 43665485-43665485 on Build GRCh38
rs752525518 Chr.19: 43665388-43665388 on Build GRCh38
rs769194446 Chr.19: 43665348-43665348 on Build GRCh38
rs745385355 Chr.19: 43665426-43665426 on Build GRCh38
rs4251833 Chr.19: 43665279-43665279 on Build GRCh38
rs987379084 Chr.19: 43665461-43665461 on Build GRCh38
rs763180998 Chr.19: 43665451-43665451 on Build GRCh38
rs763747354 Chr.19: 43665482-43665482 on Build GRCh38
rs888302757 Chr.19: 43665291-43665291 on Build GRCh38
rs755925976 Chr.19: 43665397-43665397 on Build GRCh38
rs781297129 Chr.19: 43665317-43665318 on Build GRCh38
rs775763751 Chr.19: 43665333-43665333 on Build GRCh38
rs777626185 Chr.19: 43665310-43665310 on Build GRCh38
rs150986089 Chr.19: 43665345-43665345 on Build GRCh38
rs399145 Chr.19: 43665370-43665370 on Build GRCh38
rs372921334 Chr.19: 43665307-43665307 on Build GRCh38
rs975363588 Chr.19: 43665495-43665495 on Build GRCh38
rs756784139 Chr.19: 43665280-43665281 on Build GRCh38
rs371541255 Chr.19: 43665323-43665323 on Build GRCh38
rs998471718 Chr.19: 43665318-43665318 on Build GRCh38
rs112183701 Chr.19: 43665434-43665434 on Build GRCh38

View Full Product Details