Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144404465-144404738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOBOX
Entrez Gene ID
135935
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
OG-2, OG2, OG2X, POF5, TCAG_12042
UniGene
Hs.558628
Gene Chromosome Location
Chr.7: 144396805-144410438 on Build GRCh38
Transcripts
NM_001080413.3 XM_017011742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1154782 c.146G>A p.C49Y Chr.7: 144404620-144404620 on Build GRCh38
6548943 c.98C>A p.A33D Chr.7: 144404668-144404668 on Build GRCh38
353498 c.97G>A p.A33T Chr.7: 144404669-144404669 on Build GRCh38
6515764 c.139G>T p.G47* Chr.7: 144404627-144404627 on Build GRCh38
322063 c.91C>T p.P31S Chr.7: 144404675-144404675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781182318 Chr.7: 144404579-144404580 on Build GRCh38
rs2003479 Chr.7: 144404640-144404640 on Build GRCh38
rs531700272 Chr.7: 144404636-144404636 on Build GRCh38
rs35653437 Chr.7: 144404584-144404584 on Build GRCh38
rs764306272 Chr.7: 144404673-144404673 on Build GRCh38
rs757697634 Chr.7: 144404675-144404675 on Build GRCh38
rs770306774 Chr.7: 144404572-144404572 on Build GRCh38
rs768308740 Chr.7: 144404700-144404701 on Build GRCh38
rs547261391 Chr.7: 144404550-144404550 on Build GRCh38
rs945639518 Chr.7: 144404548-144404548 on Build GRCh38
rs758931164 Chr.7: 144404564-144404564 on Build GRCh38
rs772247190 Chr.7: 144404496-144404496 on Build GRCh38
rs370043070 Chr.7: 144404628-144404628 on Build GRCh38
rs748913586 Chr.7: 144404675-144404676 on Build GRCh38
rs757794070 Chr.7: 144404625-144404625 on Build GRCh38
rs776226303 Chr.7: 144404574-144404574 on Build GRCh38
rs751956954 Chr.7: 144404674-144404674 on Build GRCh38
rs960952412 Chr.7: 144404593-144404593 on Build GRCh38
rs367904214 Chr.7: 144404545-144404545 on Build GRCh38
rs755912961 Chr.7: 144404610-144404611 on Build GRCh38
rs964825866 Chr.7: 144404676-144404676 on Build GRCh38
rs748590746 Chr.7: 144404556-144404556 on Build GRCh38
rs765614989 Chr.7: 144404605-144404605 on Build GRCh38
rs115206969 Chr.7: 144404635-144404635 on Build GRCh38
rs755658484 Chr.7: 144404689-144404689 on Build GRCh38
rs746496101 Chr.7: 144404570-144404570 on Build GRCh38
rs752926479 Chr.7: 144404649-144404649 on Build GRCh38
rs189548165 Chr.7: 144404688-144404688 on Build GRCh38
rs755267567 Chr.7: 144404551-144404551 on Build GRCh38
rs376059103 Chr.7: 144404692-144404692 on Build GRCh38
rs536180408 Chr.7: 144404690-144404690 on Build GRCh38
rs759835089 Chr.7: 144404602-144404602 on Build GRCh38
rs778571313 Chr.7: 144404704-144404704 on Build GRCh38
rs768362454 Chr.7: 144404693-144404693 on Build GRCh38
rs763258306 Chr.7: 144404670-144404670 on Build GRCh38
rs765526145 Chr.7: 144404645-144404645 on Build GRCh38
rs1017715680 Chr.7: 144404529-144404529 on Build GRCh38
rs764961142 Chr.7: 144404533-144404533 on Build GRCh38
rs200856683 Chr.7: 144404607-144404607 on Build GRCh38
rs780611966 Chr.7: 144404627-144404627 on Build GRCh38
rs750240819 Chr.7: 144404598-144404600 on Build GRCh38
rs759216900 Chr.7: 144404518-144404518 on Build GRCh38
rs762791118 Chr.7: 144404583-144404583 on Build GRCh38
rs751961900 Chr.7: 144404616-144404616 on Build GRCh38
rs371511426 Chr.7: 144404549-144404549 on Build GRCh38
rs778193142 Chr.7: 144404567-144404567 on Build GRCh38
rs781705054 Chr.7: 144404686-144404686 on Build GRCh38
rs761837506 Chr.7: 144404590-144404590 on Build GRCh38
rs766314965 Chr.7: 144404591-144404591 on Build GRCh38
rs764001433 Chr.7: 144404538-144404538 on Build GRCh38
rs775275542 Chr.7: 144404534-144404534 on Build GRCh38
rs756193964 Chr.7: 144404546-144404546 on Build GRCh38
rs140470667 Chr.7: 144404634-144404634 on Build GRCh38
rs779608853 Chr.7: 144404651-144404659 on Build GRCh38
rs753974459 Chr.7: 144404601-144404601 on Build GRCh38
rs984973208 Chr.7: 144404631-144404631 on Build GRCh38
rs745463679 Chr.7: 144404578-144404578 on Build GRCh38
rs375181430 Chr.7: 144404552-144404552 on Build GRCh38
rs768289751 Chr.7: 144404630-144404630 on Build GRCh38
rs775258475 Chr.7: 144404581-144404581 on Build GRCh38
rs758730028 Chr.7: 144404606-144404606 on Build GRCh38
rs762807681 Chr.7: 144404535-144404535 on Build GRCh38
rs769439095 Chr.7: 144404580-144404580 on Build GRCh38
rs987939830 Chr.7: 144404515-144404515 on Build GRCh38
rs757118061 Chr.7: 144404513-144404513 on Build GRCh38

View Full Product Details