Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56314804-56315065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY2
Entrez Gene ID
10330
Gene Name
canopy FGF signaling regulator 2
Gene Aliases
HP10390, MSAP, TMEM4, ZSIG9
UniGene
Hs.8752
Gene Chromosome Location
Chr.12: 56310428-56316344 on Build GRCh38
Transcripts
NM_001190991.1 NM_014255.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293118 c.113T>C p.L38P Chr.12: 56314942-56314942 on Build GRCh38
3812577 c.135G>T p.V45V Chr.12: 56314920-56314920 on Build GRCh38
6578490 c.127G>A p.A43T Chr.12: 56314928-56314928 on Build GRCh38
5005426 c.180A>G p.P60P Chr.12: 56314875-56314875 on Build GRCh38
5361695 c.167T>C p.F56S Chr.12: 56314888-56314888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527652986 Chr.12: 56314927-56314927 on Build GRCh38
rs771652866 Chr.12: 56314828-56314828 on Build GRCh38
rs764708852 Chr.12: 56315038-56315038 on Build GRCh38
rs780416784 Chr.12: 56315010-56315010 on Build GRCh38
rs749653033 Chr.12: 56315016-56315016 on Build GRCh38
rs946727184 Chr.12: 56314989-56314989 on Build GRCh38
rs1040935998 Chr.12: 56315028-56315028 on Build GRCh38
rs776150200 Chr.12: 56314908-56314908 on Build GRCh38
rs11551983 Chr.12: 56314925-56314925 on Build GRCh38
rs774485842 Chr.12: 56314909-56314909 on Build GRCh38
rs746344729 Chr.12: 56314840-56314840 on Build GRCh38
rs376920667 Chr.12: 56314838-56314838 on Build GRCh38
rs763289877 Chr.12: 56314887-56314887 on Build GRCh38
rs574865113 Chr.12: 56314936-56314936 on Build GRCh38
rs202064219 Chr.12: 56315007-56315007 on Build GRCh38
rs986121334 Chr.12: 56314999-56314999 on Build GRCh38
rs768928573 Chr.12: 56314898-56314898 on Build GRCh38
rs372058833 Chr.12: 56314930-56314930 on Build GRCh38
rs777381712 Chr.12: 56315001-56315001 on Build GRCh38
rs200294112 Chr.12: 56314941-56314941 on Build GRCh38
rs142943697 Chr.12: 56315027-56315027 on Build GRCh38
rs915291194 Chr.12: 56314979-56314979 on Build GRCh38
rs199831088 Chr.12: 56314943-56314943 on Build GRCh38
rs764996007 Chr.12: 56314948-56314948 on Build GRCh38
rs758014085 Chr.12: 56314970-56314970 on Build GRCh38
rs79111613 Chr.12: 56314922-56314922 on Build GRCh38
rs766217104 Chr.12: 56314939-56314939 on Build GRCh38
rs752350174 Chr.12: 56314961-56314961 on Build GRCh38
rs910542167 Chr.12: 56315003-56315003 on Build GRCh38
rs754753878 Chr.12: 56314945-56314945 on Build GRCh38
rs770167974 Chr.12: 56314846-56314846 on Build GRCh38
rs1017249603 Chr.12: 56314972-56314972 on Build GRCh38

View Full Product Details