Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33008504-33008775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB2
Entrez Gene ID
80341
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2
UniGene
Hs.257045
Gene Chromosome Location
Chr.20: 33007578-33023709 on Build GRCh38
Transcripts
NM_025227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2889817 c.82C>T p.R28* Chr.20: 33008656-33008656 on Build GRCh38
3758516 c.92A>T p.K31M Chr.20: 33008666-33008666 on Build GRCh38
3423585 c.54C>T p.V18V Chr.20: 33008628-33008628 on Build GRCh38
3405022 c.68C>T p.P23L Chr.20: 33008642-33008642 on Build GRCh38
7357655 c.50T>A p.V17E Chr.20: 33008624-33008624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038105186 Chr.20: 33008642-33008642 on Build GRCh38
rs764443200 Chr.20: 33008578-33008578 on Build GRCh38
rs548999891 Chr.20: 33008664-33008664 on Build GRCh38
rs373527040 Chr.20: 33008622-33008622 on Build GRCh38
rs376460103 Chr.20: 33008639-33008639 on Build GRCh38
rs967115799 Chr.20: 33008748-33008748 on Build GRCh38
rs766441754 Chr.20: 33008674-33008674 on Build GRCh38
rs373825975 Chr.20: 33008636-33008636 on Build GRCh38
rs543048762 Chr.20: 33008596-33008596 on Build GRCh38
rs201501274 Chr.20: 33008650-33008650 on Build GRCh38
rs181110734 Chr.20: 33008657-33008657 on Build GRCh38
rs979476629 Chr.20: 33008598-33008598 on Build GRCh38
rs200818769 Chr.20: 33008729-33008729 on Build GRCh38
rs746736413 Chr.20: 33008713-33008713 on Build GRCh38
rs745514889 Chr.20: 33008735-33008735 on Build GRCh38
rs1040517563 Chr.20: 33008630-33008630 on Build GRCh38
rs776610928 Chr.20: 33008564-33008564 on Build GRCh38
rs754962460 Chr.20: 33008683-33008683 on Build GRCh38
rs926925191 Chr.20: 33008584-33008584 on Build GRCh38
rs370014963 Chr.20: 33008645-33008645 on Build GRCh38
rs184795371 Chr.20: 33008747-33008747 on Build GRCh38
rs968144708 Chr.20: 33008581-33008581 on Build GRCh38
rs143951523 Chr.20: 33008626-33008626 on Build GRCh38
rs758404967 Chr.20: 33008702-33008702 on Build GRCh38
rs775991757 Chr.20: 33008569-33008569 on Build GRCh38
rs752863141 Chr.20: 33008699-33008699 on Build GRCh38
rs1049383724 Chr.20: 33008633-33008633 on Build GRCh38
rs760381008 Chr.20: 33008558-33008558 on Build GRCh38
rs143361589 Chr.20: 33008656-33008656 on Build GRCh38
rs6088067 Chr.20: 33008743-33008743 on Build GRCh38
rs6088066 Chr.20: 33008666-33008666 on Build GRCh38
rs144598883 Chr.20: 33008611-33008611 on Build GRCh38
rs373872134 Chr.20: 33008649-33008649 on Build GRCh38
rs770882531 Chr.20: 33008728-33008728 on Build GRCh38
rs80272800 Chr.20: 33008703-33008703 on Build GRCh38
rs17123986 Chr.20: 33008753-33008753 on Build GRCh38
rs149889663 Chr.20: 33008624-33008624 on Build GRCh38
rs752142338 Chr.20: 33008592-33008592 on Build GRCh38
rs140278132 Chr.20: 33008655-33008655 on Build GRCh38
rs994413193 Chr.20: 33008669-33008669 on Build GRCh38
rs565493383 Chr.20: 33008629-33008629 on Build GRCh38
rs1034002108 Chr.20: 33008691-33008691 on Build GRCh38
rs750831030 Chr.20: 33008613-33008613 on Build GRCh38
rs367764503 Chr.20: 33008533-33008533 on Build GRCh38
rs553810301 Chr.20: 33008532-33008532 on Build GRCh38
rs139467510 Chr.20: 33008574-33008574 on Build GRCh38
rs749266501 Chr.20: 33008623-33008623 on Build GRCh38
rs148646155 Chr.20: 33008689-33008689 on Build GRCh38
rs774356319 Chr.20: 33008525-33008525 on Build GRCh38
rs747828974 Chr.20: 33008542-33008542 on Build GRCh38
rs772022878 Chr.20: 33008551-33008551 on Build GRCh38
rs374709566 Chr.20: 33008632-33008632 on Build GRCh38
rs538164322 Chr.20: 33008682-33008682 on Build GRCh38
rs971312134 Chr.20: 33008553-33008553 on Build GRCh38
rs369910966 Chr.20: 33008561-33008561 on Build GRCh38
rs756321277 Chr.20: 33008615-33008615 on Build GRCh38
rs772949676 Chr.20: 33008552-33008552 on Build GRCh38

View Full Product Details