Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15676937-15677184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
UniGene
Hs.335239
Gene Chromosome Location
Chr.3: 15667236-15859814 on Build GRCh38
Transcripts
NM_001195098.1 NM_001195099.1 NM_015199.3 XM_005264996.3 XM_011533540.2 XM_011533541.2 XM_011533542.2 XM_011533543.2 XM_011533545.2 XM_011533546.2 XM_011533547.2 XM_017006026.1 XM_017006027.1 XM_017006029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059454 c.1112C>A p.T371N Chr.3: 15677046-15677046 on Build GRCh38
4770788 c.2882+1G>T p.? Chr.3: 15676973-15676973 on Build GRCh38
5863821 c.2869G>A p.A957T Chr.3: 15676987-15676987 on Build GRCh38
6600592 c.2835G>T p.K945N Chr.3: 15677021-15677021 on Build GRCh38
6996303 c.1133A>C p.E378A Chr.3: 15677025-15677025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755489229 Chr.3: 15677025-15677025 on Build GRCh38
rs374417222 Chr.3: 15677030-15677030 on Build GRCh38
rs370858006 Chr.3: 15677104-15677104 on Build GRCh38
rs779919230 Chr.3: 15677100-15677100 on Build GRCh38
rs375834695 Chr.3: 15677029-15677029 on Build GRCh38
rs148189292 Chr.3: 15677155-15677155 on Build GRCh38
rs149481554 Chr.3: 15676987-15676987 on Build GRCh38
rs2470540 Chr.3: 15677145-15677145 on Build GRCh38
rs763344003 Chr.3: 15676994-15676994 on Build GRCh38
rs563607317 Chr.3: 15676988-15676988 on Build GRCh38
rs201334606 Chr.3: 15677000-15677000 on Build GRCh38
rs143967220 Chr.3: 15676998-15676998 on Build GRCh38
rs1038778209 Chr.3: 15677113-15677113 on Build GRCh38
rs776178856 Chr.3: 15676976-15676976 on Build GRCh38
rs562323056 Chr.3: 15677062-15677062 on Build GRCh38
rs775161986 Chr.3: 15677036-15677036 on Build GRCh38
rs749512053 Chr.3: 15677079-15677079 on Build GRCh38
rs377593632 Chr.3: 15677080-15677080 on Build GRCh38
rs761298767 Chr.3: 15676984-15676984 on Build GRCh38
rs749250771 Chr.3: 15677073-15677074 on Build GRCh38
rs973722986 Chr.3: 15677041-15677041 on Build GRCh38
rs777914019 Chr.3: 15677072-15677072 on Build GRCh38
rs368294336 Chr.3: 15677039-15677039 on Build GRCh38
rs756243817 Chr.3: 15677070-15677070 on Build GRCh38
rs998010675 Chr.3: 15676983-15676983 on Build GRCh38
rs767999328 Chr.3: 15676974-15676974 on Build GRCh38

View Full Product Details