Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50839611-50839836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WFIKKN2
Entrez Gene ID
124857
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2
Gene Aliases
GASP-1, WFDC20B, WFIKKNRP, hGASP-1
UniGene
Hs.211475
Gene Chromosome Location
Chr.17: 50834650-50842353 on Build GRCh38
Transcripts
NM_175575.5 XM_017024172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3192999 c.424C>T p.P142S Chr.17: 50839712-50839712 on Build GRCh38
4579855 c.483C>T p.A161A Chr.17: 50839771-50839771 on Build GRCh38
286530 c.375C>A p.I125I Chr.17: 50839663-50839663 on Build GRCh38
6250809 c.415G>T p.E139* Chr.17: 50839703-50839703 on Build GRCh38
706874 c.544C>A p.P182T Chr.17: 50839832-50839832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779578269 Chr.17: 50839656-50839656 on Build GRCh38
rs751057329 Chr.17: 50839808-50839808 on Build GRCh38
rs754400756 Chr.17: 50839795-50839795 on Build GRCh38
rs548236113 Chr.17: 50839670-50839670 on Build GRCh38
rs147530119 Chr.17: 50839678-50839678 on Build GRCh38
rs749302087 Chr.17: 50839816-50839816 on Build GRCh38
rs771064379 Chr.17: 50839690-50839690 on Build GRCh38
rs751237590 Chr.17: 50839735-50839735 on Build GRCh38
rs759730733 Chr.17: 50839774-50839774 on Build GRCh38
rs746616318 Chr.17: 50839662-50839662 on Build GRCh38
rs201503900 Chr.17: 50839790-50839790 on Build GRCh38
rs773233764 Chr.17: 50839683-50839683 on Build GRCh38
rs574423328 Chr.17: 50839802-50839802 on Build GRCh38
rs777694304 Chr.17: 50839815-50839815 on Build GRCh38
rs764619543 Chr.17: 50839725-50839725 on Build GRCh38
rs1037925951 Chr.17: 50839659-50839659 on Build GRCh38
rs546698855 Chr.17: 50839679-50839679 on Build GRCh38
rs752327614 Chr.17: 50839813-50839813 on Build GRCh38
rs753086954 Chr.17: 50839722-50839722 on Build GRCh38
rs745407777 Chr.17: 50839637-50839637 on Build GRCh38
rs777854021 Chr.17: 50839755-50839755 on Build GRCh38
rs140159029 Chr.17: 50839726-50839726 on Build GRCh38
rs903414878 Chr.17: 50839732-50839732 on Build GRCh38
rs761033915 Chr.17: 50839785-50839785 on Build GRCh38
rs760982642 Chr.17: 50839723-50839723 on Build GRCh38
rs368900996 Chr.17: 50839731-50839731 on Build GRCh38
rs753389911 Chr.17: 50839809-50839809 on Build GRCh38
rs772363912 Chr.17: 50839775-50839775 on Build GRCh38
rs755983785 Chr.17: 50839814-50839814 on Build GRCh38
rs553049318 Chr.17: 50839771-50839771 on Build GRCh38
rs78361090 Chr.17: 50839651-50839651 on Build GRCh38
rs767816747 Chr.17: 50839698-50839698 on Build GRCh38
rs991730280 Chr.17: 50839727-50839727 on Build GRCh38
rs769670406 Chr.17: 50839748-50839748 on Build GRCh38
rs757919214 Chr.17: 50839729-50839729 on Build GRCh38
rs774446941 Chr.17: 50839695-50839695 on Build GRCh38
rs372428466 Chr.17: 50839754-50839754 on Build GRCh38
rs759787370 Chr.17: 50839697-50839697 on Build GRCh38
rs745565549 Chr.17: 50839811-50839811 on Build GRCh38
rs754627583 Chr.17: 50839736-50839736 on Build GRCh38
rs780897175 Chr.17: 50839743-50839743 on Build GRCh38
rs774462465 Chr.17: 50839772-50839772 on Build GRCh38
rs377361970 Chr.17: 50839687-50839687 on Build GRCh38
rs768393898 Chr.17: 50839668-50839668 on Build GRCh38
rs535856714 Chr.17: 50839746-50839746 on Build GRCh38
rs762303851 Chr.17: 50839801-50839801 on Build GRCh38
rs775538154 Chr.17: 50839776-50839776 on Build GRCh38
rs749177471 Chr.17: 50839759-50839759 on Build GRCh38

View Full Product Details