Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 71467165-71467436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDP1
Entrez Gene ID
55814
Gene Name
B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB
Gene Aliases
HSA238520, TAF3B1, TFC5, TFIIIB'', TFIIIB150, TFIIIB90, TFNR
UniGene
Hs.258272
Gene Chromosome Location
Chr.5: 71455615-71578288 on Build GRCh38
Transcripts
NM_018429.2 XM_011543511.2 XM_011543512.2 XM_017009630.1 XM_017009631.1 XM_017009632.1 XM_017009633.1 XM_017009634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7155154 c.860C>G p.S287C Chr.5: 71467428-71467428 on Build GRCh38
6611177 c.810delA p.G272fs*42 Chr.5: 71467378-71467378 on Build GRCh38
1069762 c.832G>T p.E278* Chr.5: 71467400-71467400 on Build GRCh38
6848265 c.840delC p.I282fs*32 Chr.5: 71467408-71467408 on Build GRCh38
6611187 c.856G>A p.G286S Chr.5: 71467424-71467424 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933600692 Chr.5: 71467290-71467290 on Build GRCh38
rs7442961 Chr.5: 71467261-71467261 on Build GRCh38
rs906822156 Chr.5: 71467400-71467400 on Build GRCh38
rs200943312 Chr.5: 71467366-71467366 on Build GRCh38
rs767611882 Chr.5: 71467279-71467279 on Build GRCh38
rs983485858 Chr.5: 71467252-71467252 on Build GRCh38
rs910557953 Chr.5: 71467326-71467326 on Build GRCh38
rs755204096 Chr.5: 71467388-71467388 on Build GRCh38
rs766423723 Chr.5: 71467214-71467214 on Build GRCh38
rs868312550 Chr.5: 71467283-71467283 on Build GRCh38
rs774625815 Chr.5: 71467358-71467358 on Build GRCh38
rs765469637 Chr.5: 71467390-71467390 on Build GRCh38
rs370946203 Chr.5: 71467403-71467403 on Build GRCh38
rs911849029 Chr.5: 71467260-71467260 on Build GRCh38
rs370553071 Chr.5: 71467404-71467404 on Build GRCh38
rs995457215 Chr.5: 71467204-71467204 on Build GRCh38
rs913501371 Chr.5: 71467347-71467347 on Build GRCh38
rs200136581 Chr.5: 71467322-71467322 on Build GRCh38
rs756544917 Chr.5: 71467245-71467245 on Build GRCh38
rs6452722 Chr.5: 71467232-71467232 on Build GRCh38
rs762235049 Chr.5: 71467360-71467360 on Build GRCh38
rs570029100 Chr.5: 71467377-71467377 on Build GRCh38
rs773800544 Chr.5: 71467367-71467367 on Build GRCh38
rs781118850 Chr.5: 71467402-71467402 on Build GRCh38
rs558177927 Chr.5: 71467327-71467327 on Build GRCh38
rs180764732 Chr.5: 71467297-71467297 on Build GRCh38
rs1025548523 Chr.5: 71467220-71467220 on Build GRCh38
rs150763040 Chr.5: 71467391-71467391 on Build GRCh38
rs753941593 Chr.5: 71467384-71467384 on Build GRCh38
rs186106001 Chr.5: 71467393-71467393 on Build GRCh38
rs760846637 Chr.5: 71467376-71467376 on Build GRCh38
rs990383313 Chr.5: 71467300-71467300 on Build GRCh38
rs972250313 Chr.5: 71467280-71467280 on Build GRCh38

View Full Product Details