Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 80462040-80462295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT3
Entrez Gene ID
346562
Gene Name
G protein subunit alpha transducin 3
Gene Aliases
GDCA
UniGene
Hs.335049
Gene Chromosome Location
Chr.7: 80457291-80512503 on Build GRCh38
Transcripts
NM_001102386.2 XM_011516124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1452199 c.814G>A p.D272N Chr.7: 80462219-80462219 on Build GRCh38
4805182 c.773A>G p.Y258C Chr.7: 80462260-80462260 on Build GRCh38
5809918 c.847C>G p.L283V Chr.7: 80462186-80462186 on Build GRCh38
6178767 c.745C>G p.L249V Chr.7: 80462288-80462288 on Build GRCh38
3412318 c.801C>G p.F267L Chr.7: 80462232-80462232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372732114 Chr.7: 80462231-80462231 on Build GRCh38
rs368632002 Chr.7: 80462201-80462201 on Build GRCh38
rs200076914 Chr.7: 80462133-80462133 on Build GRCh38
rs773661516 Chr.7: 80462219-80462220 on Build GRCh38
rs939987382 Chr.7: 80462219-80462219 on Build GRCh38
rs1047610066 Chr.7: 80462178-80462178 on Build GRCh38
rs186804021 Chr.7: 80462227-80462227 on Build GRCh38
rs771820004 Chr.7: 80462236-80462236 on Build GRCh38
rs777312113 Chr.7: 80462260-80462260 on Build GRCh38
rs936412800 Chr.7: 80462183-80462183 on Build GRCh38
rs1044709047 Chr.7: 80462153-80462153 on Build GRCh38
rs754230307 Chr.7: 80462120-80462120 on Build GRCh38
rs933173816 Chr.7: 80462177-80462177 on Build GRCh38
rs764461871 Chr.7: 80462247-80462247 on Build GRCh38
rs992461448 Chr.7: 80462076-80462076 on Build GRCh38
rs749041022 Chr.7: 80462205-80462205 on Build GRCh38
rs141648872 Chr.7: 80462262-80462262 on Build GRCh38
rs974927633 Chr.7: 80462137-80462137 on Build GRCh38
rs758450603 Chr.7: 80462154-80462154 on Build GRCh38
rs770408517 Chr.7: 80462172-80462173 on Build GRCh38
rs776813050 Chr.7: 80462086-80462086 on Build GRCh38
rs750683403 Chr.7: 80462134-80462134 on Build GRCh38
rs747181121 Chr.7: 80462193-80462193 on Build GRCh38
rs760278104 Chr.7: 80462244-80462244 on Build GRCh38
rs770717261 Chr.7: 80462215-80462215 on Build GRCh38
rs779985393 Chr.7: 80462157-80462157 on Build GRCh38
rs571029646 Chr.7: 80462146-80462146 on Build GRCh38
rs774828440 Chr.7: 80462242-80462242 on Build GRCh38

View Full Product Details