Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39200413-39200666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCCRP1
Entrez Gene ID
342897
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
FBXO50, NCCRP-1
UniGene
Hs.726934
Gene Chromosome Location
Chr.19: 39196964-39201882 on Build GRCh38
Transcripts
NM_001001414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4077915 c.719G>A p.G240D Chr.19: 39200647-39200647 on Build GRCh38
3223421 c.640G>A p.V214M Chr.19: 39200437-39200437 on Build GRCh38
3223424 c.677G>A p.R226H Chr.19: 39200474-39200474 on Build GRCh38
6479265 c.718G>C p.G240R Chr.19: 39200646-39200646 on Build GRCh38
4404817 c.656C>T p.S219F Chr.19: 39200453-39200453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752220471 Chr.19: 39200484-39200484 on Build GRCh38
rs751063241 Chr.19: 39200533-39200533 on Build GRCh38
rs749566718 Chr.19: 39200442-39200442 on Build GRCh38
rs773311053 Chr.19: 39200505-39200505 on Build GRCh38
rs746170004 Chr.19: 39200541-39200541 on Build GRCh38
rs140808558 Chr.19: 39200568-39200568 on Build GRCh38
rs757283881 Chr.19: 39200646-39200646 on Build GRCh38
rs747046037 Chr.19: 39200586-39200586 on Build GRCh38
rs201299839 Chr.19: 39200474-39200474 on Build GRCh38
rs376755212 Chr.19: 39200613-39200613 on Build GRCh38
rs781496915 Chr.19: 39200436-39200436 on Build GRCh38
rs755253469 Chr.19: 39200499-39200499 on Build GRCh38
rs370150617 Chr.19: 39200521-39200521 on Build GRCh38
rs750937492 Chr.19: 39200495-39200495 on Build GRCh38
rs749663501 Chr.19: 39200498-39200498 on Build GRCh38
rs763556429 Chr.19: 39200525-39200525 on Build GRCh38
rs746270847 Chr.19: 39200437-39200437 on Build GRCh38
rs202157094 Chr.19: 39200598-39200598 on Build GRCh38
rs757809951 Chr.19: 39200479-39200479 on Build GRCh38
rs550666456 Chr.19: 39200566-39200566 on Build GRCh38
rs554359821 Chr.19: 39200502-39200502 on Build GRCh38
rs777387736 Chr.19: 39200492-39200492 on Build GRCh38
rs11672471 Chr.19: 39200496-39200496 on Build GRCh38
rs746977351 Chr.19: 39200506-39200506 on Build GRCh38
rs1011855765 Chr.19: 39200560-39200560 on Build GRCh38
rs371735275 Chr.19: 39200645-39200645 on Build GRCh38
rs767016876 Chr.19: 39200619-39200619 on Build GRCh38
rs776505034 Chr.19: 39200591-39200591 on Build GRCh38
rs772005313 Chr.19: 39200504-39200504 on Build GRCh38
rs775337353 Chr.19: 39200603-39200603 on Build GRCh38
rs776417815 Chr.19: 39200510-39200510 on Build GRCh38
rs774636871 Chr.19: 39200451-39200451 on Build GRCh38
rs749536977 Chr.19: 39200630-39200632 on Build GRCh38
rs770852145 Chr.19: 39200507-39200507 on Build GRCh38
rs780196086 Chr.19: 39200584-39200585 on Build GRCh38
rs1056153693 Chr.19: 39200514-39200514 on Build GRCh38
rs535604747 Chr.19: 39200520-39200520 on Build GRCh38
rs367555888 Chr.19: 39200616-39200616 on Build GRCh38
rs759253440 Chr.19: 39200473-39200473 on Build GRCh38
rs781688368 Chr.19: 39200501-39200501 on Build GRCh38
rs768983205 Chr.19: 39200447-39200447 on Build GRCh38
rs763794783 Chr.19: 39200608-39200608 on Build GRCh38
rs138080176 Chr.19: 39200455-39200455 on Build GRCh38
rs757801641 Chr.19: 39200485-39200485 on Build GRCh38
rs760549795 Chr.19: 39200462-39200462 on Build GRCh38
rs753617780 Chr.19: 39200466-39200466 on Build GRCh38
rs78285559 Chr.19: 39200567-39200567 on Build GRCh38
rs772059052 Chr.19: 39200454-39200454 on Build GRCh38
rs376147535 Chr.19: 39200516-39200516 on Build GRCh38
rs770870612 Chr.19: 39200590-39200590 on Build GRCh38
rs968503038 Chr.19: 39200481-39200481 on Build GRCh38
rs766924485 Chr.19: 39200534-39200534 on Build GRCh38
rs756352900 Chr.19: 39200440-39200441 on Build GRCh38
rs754358994 Chr.19: 39200624-39200624 on Build GRCh38
rs191938103 Chr.19: 39200453-39200453 on Build GRCh38
rs1044410884 Chr.19: 39200561-39200561 on Build GRCh38
rs11672475 Chr.19: 39200576-39200576 on Build GRCh38
rs754266995 Chr.19: 39200565-39200565 on Build GRCh38
rs766210483 Chr.19: 39200465-39200465 on Build GRCh38
rs375697876 Chr.19: 39200440-39200440 on Build GRCh38
rs367877232 Chr.19: 39200625-39200625 on Build GRCh38
rs765570355 Chr.19: 39200631-39200631 on Build GRCh38
rs762567731 Chr.19: 39200604-39200604 on Build GRCh38
rs11672479 Chr.19: 39200599-39200599 on Build GRCh38
rs774535300 Chr.19: 39200632-39200632 on Build GRCh38
rs377645422 Chr.19: 39200643-39200643 on Build GRCh38
rs144574051 Chr.19: 39200577-39200577 on Build GRCh38

View Full Product Details