Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 84152449-84152710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP162
Entrez Gene ID
22832
Gene Name
centrosomal protein 162
Gene Aliases
C6orf84, KIAA1009, QN1
UniGene
Hs.485865
Gene Chromosome Location
Chr.6: 84124241-84227889 on Build GRCh38
Transcripts
NM_001286206.1 NM_014895.3 XM_005248674.3 XM_006715380.2 XM_011535592.2 XM_017010483.1 XM_017010484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4570973 c.3578T>G p.L1193R Chr.6: 84152596-84152596 on Build GRCh38
6442245 c.3500C>T p.S1167F Chr.6: 84152674-84152674 on Build GRCh38
6024300 c.3541_3542insA p.N1181fs*2 Chr.6: 84152632-84152633 on Build GRCh38
673702 c.3559A>G p.I1187V Chr.6: 84152615-84152615 on Build GRCh38
5939543 c.3509C>T p.S1170L Chr.6: 84152665-84152665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042129092 Chr.6: 84152547-84152547 on Build GRCh38
rs925379309 Chr.6: 84152543-84152543 on Build GRCh38
rs181627156 Chr.6: 84152548-84152548 on Build GRCh38
rs756490203 Chr.6: 84152623-84152623 on Build GRCh38
rs757580659 Chr.6: 84152654-84152654 on Build GRCh38
rs769554715 Chr.6: 84152679-84152679 on Build GRCh38
rs773029329 Chr.6: 84152682-84152682 on Build GRCh38
rs754802254 Chr.6: 84152668-84152668 on Build GRCh38
rs1009736246 Chr.6: 84152522-84152522 on Build GRCh38
rs745875869 Chr.6: 84152593-84152597 on Build GRCh38
rs955362311 Chr.6: 84152664-84152664 on Build GRCh38
rs745971907 Chr.6: 84152661-84152661 on Build GRCh38
rs143512455 Chr.6: 84152598-84152598 on Build GRCh38
rs753214927 Chr.6: 84152610-84152610 on Build GRCh38
rs751103877 Chr.6: 84152556-84152556 on Build GRCh38
rs757412911 Chr.6: 84152503-84152503 on Build GRCh38
rs577906341 Chr.6: 84152541-84152541 on Build GRCh38
rs770584366 Chr.6: 84152689-84152689 on Build GRCh38
rs754085371 Chr.6: 84152645-84152645 on Build GRCh38
rs1017429203 Chr.6: 84152573-84152573 on Build GRCh38
rs773464642 Chr.6: 84152642-84152642 on Build GRCh38
rs779083268 Chr.6: 84152657-84152657 on Build GRCh38
rs748091431 Chr.6: 84152670-84152670 on Build GRCh38
rs1052406998 Chr.6: 84152627-84152627 on Build GRCh38
rs748217597 Chr.6: 84152481-84152481 on Build GRCh38
rs761126214 Chr.6: 84152542-84152542 on Build GRCh38
rs903595196 Chr.6: 84152554-84152554 on Build GRCh38
rs781184016 Chr.6: 84152669-84152669 on Build GRCh38
rs368672446 Chr.6: 84152630-84152630 on Build GRCh38
rs781387307 Chr.6: 84152529-84152533 on Build GRCh38
rs111956724 Chr.6: 84152593-84152593 on Build GRCh38
rs560842687 Chr.6: 84152578-84152578 on Build GRCh38
rs748891806 Chr.6: 84152683-84152683 on Build GRCh38

View Full Product Details