Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53073436-53073709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO9
Entrez Gene ID
26268
Gene Name
F-box protein 9
Gene Aliases
FBX9, NY-REN-57, VCIA1, dJ341E18.2
UniGene
Hs.216653
Gene Chromosome Location
Chr.6: 53057807-53100873 on Build GRCh38
Transcripts
NM_012347.4 NM_033480.2 NM_033481.3 XM_005248995.4 XM_005248996.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1080303 c.129C>A p.L43L Chr.6: 53073489-53073489 on Build GRCh38
1080305 c.140G>A p.R47Q Chr.6: 53073500-53073500 on Build GRCh38
3355276 c.236C>T p.S79L Chr.6: 53073596-53073596 on Build GRCh38
218888 c.221C>T p.S74F Chr.6: 53073581-53073581 on Build GRCh38
3662625 c.236C>A p.S79* Chr.6: 53073596-53073596 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533226016 Chr.6: 53073605-53073605 on Build GRCh38
rs756294442 Chr.6: 53073591-53073591 on Build GRCh38
rs764778954 Chr.6: 53073469-53073469 on Build GRCh38
rs758167975 Chr.6: 53073671-53073673 on Build GRCh38
rs761265862 Chr.6: 53073459-53073459 on Build GRCh38
rs898970767 Chr.6: 53073606-53073606 on Build GRCh38
rs771154563 Chr.6: 53073596-53073596 on Build GRCh38
rs950935201 Chr.6: 53073478-53073478 on Build GRCh38
rs866942317 Chr.6: 53073532-53073532 on Build GRCh38
rs761228474 Chr.6: 53073624-53073624 on Build GRCh38
rs766065276 Chr.6: 53073495-53073495 on Build GRCh38
rs772855482 Chr.6: 53073644-53073644 on Build GRCh38
rs575774353 Chr.6: 53073672-53073672 on Build GRCh38
rs991285816 Chr.6: 53073562-53073562 on Build GRCh38
rs759387823 Chr.6: 53073469-53073469 on Build GRCh38
rs75345592 Chr.6: 53073672-53073672 on Build GRCh38
rs765155339 Chr.6: 53073671-53073672 on Build GRCh38
rs982171772 Chr.6: 53073483-53073483 on Build GRCh38
rs750156058 Chr.6: 53073477-53073477 on Build GRCh38
rs5876299 Chr.6: 53073671-53073671 on Build GRCh38
rs749297680 Chr.6: 53073594-53073594 on Build GRCh38
rs943150487 Chr.6: 53073658-53073658 on Build GRCh38
rs541414181 Chr.6: 53073568-53073568 on Build GRCh38
rs796682661 Chr.6: 53073683-53073683 on Build GRCh38
rs754758186 Chr.6: 53073559-53073559 on Build GRCh38
rs1051468068 Chr.6: 53073653-53073653 on Build GRCh38
rs564304814 Chr.6: 53073481-53073481 on Build GRCh38
rs978657377 Chr.6: 53073572-53073572 on Build GRCh38
rs199921575 Chr.6: 53073580-53073580 on Build GRCh38
rs751217456 Chr.6: 53073654-53073654 on Build GRCh38
rs761179184 Chr.6: 53073550-53073550 on Build GRCh38
rs375274718 Chr.6: 53073597-53073597 on Build GRCh38
rs899337920 Chr.6: 53073669-53073669 on Build GRCh38
rs777462567 Chr.6: 53073670-53073671 on Build GRCh38
rs371936172 Chr.6: 53073581-53073581 on Build GRCh38
rs766152973 Chr.6: 53073652-53073652 on Build GRCh38
rs551672134 Chr.6: 53073656-53073656 on Build GRCh38
rs772447447 Chr.6: 53073602-53073602 on Build GRCh38
rs1035415962 Chr.6: 53073499-53073499 on Build GRCh38
rs879103739 Chr.6: 53073673-53073673 on Build GRCh38
rs987422960 Chr.6: 53073468-53073468 on Build GRCh38
rs910495879 Chr.6: 53073564-53073564 on Build GRCh38
rs762496671 Chr.6: 53073650-53073650 on Build GRCh38
rs769493763 Chr.6: 53073639-53073639 on Build GRCh38
rs200223460 Chr.6: 53073684-53073684 on Build GRCh38
rs762692634 Chr.6: 53073491-53073491 on Build GRCh38
rs775906236 Chr.6: 53073614-53073614 on Build GRCh38
rs968484396 Chr.6: 53073570-53073570 on Build GRCh38
rs368555675 Chr.6: 53073661-53073661 on Build GRCh38

View Full Product Details