Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58840083-58840312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs755781835 Chr.20: 58840237-58840237 on Build GRCh38
rs141238454 Chr.20: 58840273-58840273 on Build GRCh38
rs975964450 Chr.20: 58840216-58840216 on Build GRCh38
rs767657815 Chr.20: 58840120-58840120 on Build GRCh38
rs774788451 Chr.20: 58840247-58840247 on Build GRCh38
rs759833927 Chr.20: 58840188-58840188 on Build GRCh38
rs780943650 Chr.20: 58840163-58840163 on Build GRCh38
rs755729283 Chr.20: 58840290-58840290 on Build GRCh38
rs967292772 Chr.20: 58840182-58840182 on Build GRCh38
rs746736450 Chr.20: 58840161-58840161 on Build GRCh38
rs370175837 Chr.20: 58840232-58840232 on Build GRCh38
rs760919160 Chr.20: 58840210-58840210 on Build GRCh38
rs778940007 Chr.20: 58840144-58840144 on Build GRCh38
rs767745741 Chr.20: 58840196-58840196 on Build GRCh38
rs775901436 Chr.20: 58840257-58840257 on Build GRCh38
rs1800901 Chr.20: 58840289-58840289 on Build GRCh38
rs767440362 Chr.20: 58840111-58840112 on Build GRCh38
rs754947062 Chr.20: 58840272-58840272 on Build GRCh38
rs761937008 Chr.20: 58840270-58840270 on Build GRCh38
rs558969394 Chr.20: 58840223-58840223 on Build GRCh38
rs772759887 Chr.20: 58840169-58840169 on Build GRCh38
rs780051524 Chr.20: 58840149-58840149 on Build GRCh38
rs761688848 Chr.20: 58840109-58840110 on Build GRCh38
rs764128447 Chr.20: 58840214-58840214 on Build GRCh38
rs202131370 Chr.20: 58840113-58840113 on Build GRCh38
rs774390751 Chr.20: 58840103-58840105 on Build GRCh38
rs747725720 Chr.20: 58840164-58840164 on Build GRCh38
rs777218004 Chr.20: 58840239-58840239 on Build GRCh38
rs752831442 Chr.20: 58840141-58840141 on Build GRCh38
rs374513507 Chr.20: 58840106-58840106 on Build GRCh38
rs565827536 Chr.20: 58840101-58840101 on Build GRCh38
rs144308406 Chr.20: 58840116-58840116 on Build GRCh38
rs375337259 Chr.20: 58840269-58840269 on Build GRCh38
rs139302910 Chr.20: 58840114-58840114 on Build GRCh38
rs146532795 Chr.20: 58840244-58840244 on Build GRCh38
rs747808766 Chr.20: 58840235-58840235 on Build GRCh38
rs764371346 Chr.20: 58840258-58840258 on Build GRCh38
rs752818515 Chr.20: 58840202-58840202 on Build GRCh38
rs569736160 Chr.20: 58840174-58840174 on Build GRCh38
rs772464689 Chr.20: 58840253-58840253 on Build GRCh38
rs774700482 Chr.20: 58840183-58840183 on Build GRCh38
rs112532266 Chr.20: 58840230-58840230 on Build GRCh38
rs199948732 Chr.20: 58840252-58840252 on Build GRCh38
rs548132075 Chr.20: 58840147-58840147 on Build GRCh38
rs750367565 Chr.20: 58840146-58840146 on Build GRCh38
rs1002304696 Chr.20: 58840111-58840111 on Build GRCh38
rs754780507 Chr.20: 58840234-58840234 on Build GRCh38
rs537714939 Chr.20: 58840177-58840177 on Build GRCh38
rs557714686 Chr.20: 58840242-58840242 on Build GRCh38
rs758474465 Chr.20: 58840229-58840229 on Build GRCh38
rs776589538 Chr.20: 58840260-58840260 on Build GRCh38

View Full Product Details