Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92167906-92168135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1107
Entrez Gene ID
23285
Gene Name
KIAA1107
Gene Aliases
-
UniGene
Hs.21554
Gene Chromosome Location
Chr.1: 92167052-92188509 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344891 c.103C>T p.L35L Chr.1: 92167959-92167959 on Build GRCh38
5134442 c.78_80delTCT p.L28delL Chr.1: 92167934-92167936 on Build GRCh38
7288318 c.73T>C p.C25R Chr.1: 92167929-92167929 on Build GRCh38
6460775 c.222C>T p.I74I Chr.1: 92167913-92167913 on Build GRCh38
5154457 c.268C>T p.L90L Chr.1: 92167959-92167959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369486791 Chr.1: 92167938-92167938 on Build GRCh38
rs113966725 Chr.1: 92168017-92168017 on Build GRCh38
rs919902050 Chr.1: 92168022-92168022 on Build GRCh38
rs768235701 Chr.1: 92167952-92167952 on Build GRCh38
rs919864370 Chr.1: 92168000-92168000 on Build GRCh38
rs747797798 Chr.1: 92167984-92167984 on Build GRCh38
rs969491443 Chr.1: 92168016-92168016 on Build GRCh38
rs916650440 Chr.1: 92167982-92167982 on Build GRCh38
rs780985809 Chr.1: 92168054-92168054 on Build GRCh38
rs973983538 Chr.1: 92167976-92167976 on Build GRCh38
rs1044422395 Chr.1: 92168049-92168049 on Build GRCh38
rs912842271 Chr.1: 92168073-92168073 on Build GRCh38
rs754583194 Chr.1: 92168043-92168043 on Build GRCh38
rs928336990 Chr.1: 92168060-92168060 on Build GRCh38
rs938389604 Chr.1: 92168063-92168063 on Build GRCh38
rs749733875 Chr.1: 92167944-92167944 on Build GRCh38
rs992434764 Chr.1: 92167965-92167965 on Build GRCh38
rs778659008 Chr.1: 92168021-92168021 on Build GRCh38
rs1047277839 Chr.1: 92168065-92168065 on Build GRCh38
rs960535700 Chr.1: 92167963-92167963 on Build GRCh38

View Full Product Details