Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132133467-132133700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
UniGene
Hs.106254
Gene Chromosome Location
Chr.10: 132036068-132184852 on Build GRCh38
Transcripts
NM_001105521.3 NM_001323086.1 NM_001323087.1 NM_001323088.1 NM_001323089.1 NM_001323090.1 XM_005252674.4 XM_005252676.4 XM_011539676.2 XM_011539682.2 XM_017016095.1 XM_017016096.1 XM_017016097.1 XM_017016098.1 XM_017016099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6208636 c.823G>A p.D275N Chr.10: 132133501-132133501 on Build GRCh38
4439730 c.795G>A p.R265R Chr.10: 132133473-132133473 on Build GRCh38
5783999 c.800T>G p.L267R Chr.10: 132133478-132133478 on Build GRCh38
6367913 c.835C>T p.H279Y Chr.10: 132133513-132133513 on Build GRCh38
4983771 c.793C>T p.R265W Chr.10: 132133471-132133471 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750112337 Chr.10: 132133504-132133504 on Build GRCh38
rs952283129 Chr.10: 132133613-132133613 on Build GRCh38
rs376769665 Chr.10: 132133503-132133503 on Build GRCh38
rs376847032 Chr.10: 132133562-132133562 on Build GRCh38
rs975027682 Chr.10: 132133611-132133611 on Build GRCh38
rs778936209 Chr.10: 132133551-132133551 on Build GRCh38
rs186075669 Chr.10: 132133612-132133612 on Build GRCh38
rs925365426 Chr.10: 132133599-132133599 on Build GRCh38
rs757056866 Chr.10: 132133525-132133526 on Build GRCh38
rs1056583049 Chr.10: 132133659-132133659 on Build GRCh38
rs965023236 Chr.10: 132133524-132133524 on Build GRCh38
rs998973808 Chr.10: 132133571-132133571 on Build GRCh38
rs916573425 Chr.10: 132133615-132133615 on Build GRCh38
rs373638615 Chr.10: 132133630-132133630 on Build GRCh38
rs773877735 Chr.10: 132133496-132133496 on Build GRCh38
rs372501697 Chr.10: 132133550-132133550 on Build GRCh38
rs1010735019 Chr.10: 132133559-132133559 on Build GRCh38
rs373581338 Chr.10: 132133558-132133558 on Build GRCh38
rs558332394 Chr.10: 132133517-132133517 on Build GRCh38
rs917592748 Chr.10: 132133667-132133667 on Build GRCh38
rs766327711 Chr.10: 132133518-132133518 on Build GRCh38
rs947686644 Chr.10: 132133675-132133675 on Build GRCh38
rs535654063 Chr.10: 132133634-132133634 on Build GRCh38
rs940478637 Chr.10: 132133636-132133636 on Build GRCh38
rs370000537 Chr.10: 132133557-132133557 on Build GRCh38
rs753654773 Chr.10: 132133544-132133544 on Build GRCh38
rs966408218 Chr.10: 132133578-132133578 on Build GRCh38
rs984944979 Chr.10: 132133622-132133622 on Build GRCh38

View Full Product Details