Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58249537-58249762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPO
Entrez Gene ID
4025
Gene Name
lactoperoxidase
Gene Aliases
SPO
UniGene
Hs.234742
Gene Chromosome Location
Chr.17: 58238426-58268518 on Build GRCh38
Transcripts
NM_001160102.1 NM_006151.2 NR_027647.1 XM_011524808.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4642143 c.551G>A p.R184H Chr.17: 58249673-58249673 on Build GRCh38
1384683 c.456G>A p.A152A Chr.17: 58249578-58249578 on Build GRCh38
6702627 c.452C>A p.P151H Chr.17: 58249574-58249574 on Build GRCh38
4068106 c.463G>A p.A155T Chr.17: 58249585-58249585 on Build GRCh38
1384684 c.466G>A p.A156T Chr.17: 58249588-58249588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752679144 Chr.17: 58249623-58249623 on Build GRCh38
rs774040085 Chr.17: 58249709-58249709 on Build GRCh38
rs777874385 Chr.17: 58249635-58249635 on Build GRCh38
rs903759692 Chr.17: 58249701-58249701 on Build GRCh38
rs1022724266 Chr.17: 58249733-58249733 on Build GRCh38
rs779137282 Chr.17: 58249661-58249661 on Build GRCh38
rs757007202 Chr.17: 58249643-58249643 on Build GRCh38
rs761233980 Chr.17: 58249566-58249566 on Build GRCh38
rs377627689 Chr.17: 58249692-58249692 on Build GRCh38
rs956350329 Chr.17: 58249723-58249723 on Build GRCh38
rs757060080 Chr.17: 58249730-58249730 on Build GRCh38
rs749069299 Chr.17: 58249642-58249642 on Build GRCh38
rs8178332 Chr.17: 58249665-58249665 on Build GRCh38
rs767194529 Chr.17: 58249607-58249607 on Build GRCh38
rs772131545 Chr.17: 58249670-58249670 on Build GRCh38
rs759178843 Chr.17: 58249712-58249712 on Build GRCh38
rs756162415 Chr.17: 58249633-58249633 on Build GRCh38
rs547768460 Chr.17: 58249704-58249704 on Build GRCh38
rs867375481 Chr.17: 58249593-58249593 on Build GRCh38
rs777033131 Chr.17: 58249578-58249578 on Build GRCh38
rs764054113 Chr.17: 58249728-58249728 on Build GRCh38
rs778636807 Chr.17: 58249739-58249739 on Build GRCh38
rs564325929 Chr.17: 58249570-58249570 on Build GRCh38
rs1007119280 Chr.17: 58249684-58249684 on Build GRCh38
rs762109945 Chr.17: 58249585-58249585 on Build GRCh38
rs8178333 Chr.17: 58249702-58249702 on Build GRCh38
rs751180138 Chr.17: 58249603-58249603 on Build GRCh38
rs991799383 Chr.17: 58249726-58249726 on Build GRCh38
rs746680338 Chr.17: 58249672-58249690 on Build GRCh38
rs766019233 Chr.17: 58249594-58249594 on Build GRCh38
rs752305226 Chr.17: 58249714-58249714 on Build GRCh38
rs929981757 Chr.17: 58249579-58249579 on Build GRCh38
rs1051155731 Chr.17: 58249620-58249620 on Build GRCh38
rs1012303395 Chr.17: 58249716-58249716 on Build GRCh38
rs140216571 Chr.17: 58249604-58249604 on Build GRCh38
rs755715378 Chr.17: 58249718-58249718 on Build GRCh38
rs750856669 Chr.17: 58249586-58249586 on Build GRCh38
rs369376380 Chr.17: 58249713-58249713 on Build GRCh38
rs775584287 Chr.17: 58249679-58249679 on Build GRCh38

View Full Product Details