Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138272922-138273159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_015396.5 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 NM_178130.3

Gene Symbol
NME9
Entrez Gene ID
347736
Gene Name
NME/NM23 family member 9
Gene Aliases
NM23-H9, TXL-2, TXL2, TXNDC6
UniGene
Hs.660992
Gene Chromosome Location
Chr.3: 138261437-138331785 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_015396.5 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 NM_178130.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373034 c.1453G>A p.A485T Chr.3: 138272982-138272982 on Build GRCh38
7169202 c.1624C>T p.R542C Chr.3: 138273111-138273111 on Build GRCh38
3408277 c.1518A>G p.L506L Chr.3: 138273047-138273047 on Build GRCh38
5221310 c.1540delG p.E514fs*12 Chr.3: 138273027-138273027 on Build GRCh38
5350904 c.1438-1G>A p.? Chr.3: 138272966-138272966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369744825 Chr.3: 138273017-138273017 on Build GRCh38
rs185968098 Chr.3: 138273040-138273040 on Build GRCh38
rs761193933 Chr.3: 138273131-138273131 on Build GRCh38
rs771223689 Chr.3: 138273092-138273092 on Build GRCh38
rs767890360 Chr.3: 138273107-138273107 on Build GRCh38
rs753239950 Chr.3: 138272966-138272966 on Build GRCh38
rs985393751 Chr.3: 138272980-138272980 on Build GRCh38
rs775907909 Chr.3: 138273109-138273109 on Build GRCh38
rs952963574 Chr.3: 138273038-138273038 on Build GRCh38
rs376712056 Chr.3: 138272971-138272971 on Build GRCh38
rs764780936 Chr.3: 138273000-138273000 on Build GRCh38
rs774698014 Chr.3: 138273104-138273104 on Build GRCh38
rs773305490 Chr.3: 138273077-138273077 on Build GRCh38
rs961015317 Chr.3: 138272960-138272960 on Build GRCh38
rs750096434 Chr.3: 138273015-138273015 on Build GRCh38
rs748097277 Chr.3: 138273060-138273060 on Build GRCh38
rs372033527 Chr.3: 138272970-138272970 on Build GRCh38
rs769789020 Chr.3: 138273062-138273062 on Build GRCh38
rs749529780 Chr.3: 138273081-138273081 on Build GRCh38
rs779828353 Chr.3: 138273031-138273031 on Build GRCh38
rs767964327 Chr.3: 138272959-138272959 on Build GRCh38
rs746864519 Chr.3: 138273035-138273035 on Build GRCh38
rs754763078 Chr.3: 138273039-138273039 on Build GRCh38

View Full Product Details