Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10354069-10354330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYK2
Entrez Gene ID
7297
Gene Name
tyrosine kinase 2
Gene Aliases
IMD35, JTK1
UniGene
Hs.75516
Gene Chromosome Location
Chr.19: 10350528-10380572 on Build GRCh38
Transcripts
NM_003331.4 XM_011528245.1 XM_011528246.2 XM_011528247.1 XM_011528249.2 XM_011528251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1240593 c.2855G>A p.R952H Chr.19: 10354095-10354095 on Build GRCh38
327326 c.2810G>C p.G937A Chr.19: 10354140-10354140 on Build GRCh38
327325 c.2863T>C p.Y955H Chr.19: 10354087-10354087 on Build GRCh38
4738522 c.2763C>T p.D921D Chr.19: 10354187-10354187 on Build GRCh38
6814657 c.2871G>A p.E957E Chr.19: 10354079-10354079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116502549 Chr.19: 10354273-10354273 on Build GRCh38
rs774268489 Chr.19: 10354112-10354112 on Build GRCh38
rs150832201 Chr.19: 10354306-10354306 on Build GRCh38
rs554711523 Chr.19: 10354128-10354128 on Build GRCh38
rs754351041 Chr.19: 10354107-10354107 on Build GRCh38
rs749465990 Chr.19: 10354210-10354210 on Build GRCh38
rs748715653 Chr.19: 10354199-10354200 on Build GRCh38
rs35018800 Chr.19: 10354167-10354167 on Build GRCh38
rs1003072749 Chr.19: 10354142-10354142 on Build GRCh38
rs781770609 Chr.19: 10354195-10354195 on Build GRCh38
rs775373425 Chr.19: 10354241-10354241 on Build GRCh38
rs760207885 Chr.19: 10354130-10354130 on Build GRCh38
rs751960997 Chr.19: 10354187-10354187 on Build GRCh38
rs761308551 Chr.19: 10354253-10354253 on Build GRCh38
rs963707334 Chr.19: 10354172-10354172 on Build GRCh38
rs141407156 Chr.19: 10354295-10354295 on Build GRCh38
rs763629813 Chr.19: 10354136-10354136 on Build GRCh38
rs745696767 Chr.19: 10354237-10354237 on Build GRCh38
rs948005175 Chr.19: 10354303-10354303 on Build GRCh38
rs139288588 Chr.19: 10354200-10354200 on Build GRCh38
rs138711713 Chr.19: 10354162-10354163 on Build GRCh38
rs56085935 Chr.19: 10354133-10354133 on Build GRCh38
rs779111054 Chr.19: 10354198-10354199 on Build GRCh38
rs56255228 Chr.19: 10354158-10354158 on Build GRCh38
rs756231936 Chr.19: 10354208-10354208 on Build GRCh38
rs772009258 Chr.19: 10354122-10354122 on Build GRCh38
rs954726975 Chr.19: 10354262-10354262 on Build GRCh38
rs778512258 Chr.19: 10354226-10354226 on Build GRCh38
rs56286467 Chr.19: 10354289-10354289 on Build GRCh38
rs748253254 Chr.19: 10354091-10354091 on Build GRCh38
rs192892841 Chr.19: 10354276-10354276 on Build GRCh38
rs770093652 Chr.19: 10354092-10354092 on Build GRCh38
rs749565585 Chr.19: 10354104-10354104 on Build GRCh38
rs778049174 Chr.19: 10354209-10354209 on Build GRCh38
rs12720319 Chr.19: 10354244-10354244 on Build GRCh38
rs769496322 Chr.19: 10354283-10354284 on Build GRCh38
rs755488436 Chr.19: 10354191-10354191 on Build GRCh38
rs56334302 Chr.19: 10354238-10354238 on Build GRCh38
rs772403799 Chr.19: 10354201-10354202 on Build GRCh38
rs776212121 Chr.19: 10354203-10354204 on Build GRCh38
rs759322708 Chr.19: 10354119-10354119 on Build GRCh38
rs1053309682 Chr.19: 10354145-10354145 on Build GRCh38
rs370899700 Chr.19: 10354247-10354247 on Build GRCh38
rs367715203 Chr.19: 10354121-10354121 on Build GRCh38
rs577565850 Chr.19: 10354246-10354246 on Build GRCh38
rs760012742 Chr.19: 10354284-10354284 on Build GRCh38
rs745379535 Chr.19: 10354205-10354206 on Build GRCh38
rs771188635 Chr.19: 10354216-10354216 on Build GRCh38
rs778172039 Chr.19: 10354098-10354098 on Build GRCh38

View Full Product Details