Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73527606-73527875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAU2
Entrez Gene ID
27067
Gene Name
staufen double-stranded RNA binding protein 2
Gene Aliases
39K2, 39K3
UniGene
Hs.561815
Gene Chromosome Location
Chr.8: 73420369-73747708 on Build GRCh38
Transcripts
NM_001164380.1 NM_001164381.1 NM_001164382.1 NM_001164383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6582585 c.1332+1G>A p.? Chr.8: 73527690-73527690 on Build GRCh38
486649 c.1241G>T p.S414I Chr.8: 73527782-73527782 on Build GRCh38
4776687 c.1287G>C p.M429I Chr.8: 73527736-73527736 on Build GRCh38
6530419 c.1306A>G p.S436G Chr.8: 73527717-73527717 on Build GRCh38
6013188 c.1225T>G p.Y409D Chr.8: 73527798-73527798 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370898737 Chr.8: 73527726-73527726 on Build GRCh38
rs1056356317 Chr.8: 73527775-73527776 on Build GRCh38
rs1042435119 Chr.8: 73527754-73527754 on Build GRCh38
rs1012053505 Chr.8: 73527808-73527808 on Build GRCh38
rs915299329 Chr.8: 73527711-73527711 on Build GRCh38
rs566097434 Chr.8: 73527770-73527770 on Build GRCh38
rs777791461 Chr.8: 73527801-73527801 on Build GRCh38
rs975239273 Chr.8: 73527710-73527710 on Build GRCh38
rs557886051 Chr.8: 73527847-73527847 on Build GRCh38
rs927405994 Chr.8: 73527662-73527662 on Build GRCh38
rs771119614 Chr.8: 73527840-73527840 on Build GRCh38
rs758227793 Chr.8: 73527694-73527694 on Build GRCh38
rs572469683 Chr.8: 73527698-73527698 on Build GRCh38
rs942210865 Chr.8: 73527646-73527646 on Build GRCh38
rs73326769 Chr.8: 73527647-73527647 on Build GRCh38
rs867533215 Chr.8: 73527765-73527765 on Build GRCh38
rs1057169578 Chr.8: 73527723-73527723 on Build GRCh38
rs891789520 Chr.8: 73527772-73527772 on Build GRCh38
rs761496192 Chr.8: 73527757-73527760 on Build GRCh38
rs139710481 Chr.8: 73527820-73527820 on Build GRCh38
rs943508665 Chr.8: 73527655-73527655 on Build GRCh38
rs1042338212 Chr.8: 73527791-73527791 on Build GRCh38
rs894990037 Chr.8: 73527806-73527806 on Build GRCh38
rs902503376 Chr.8: 73527763-73527763 on Build GRCh38
rs756385869 Chr.8: 73527731-73527731 on Build GRCh38
rs765219622 Chr.8: 73527661-73527661 on Build GRCh38
rs868188270 Chr.8: 73527780-73527780 on Build GRCh38
rs376807384 Chr.8: 73527712-73527712 on Build GRCh38
rs779799752 Chr.8: 73527724-73527724 on Build GRCh38
rs912099426 Chr.8: 73527652-73527652 on Build GRCh38
rs750358214 Chr.8: 73527675-73527675 on Build GRCh38
rs753397504 Chr.8: 73527641-73527641 on Build GRCh38
rs1010200856 Chr.8: 73527788-73527788 on Build GRCh38

View Full Product Details