Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110137073-110137312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H12C
Entrez Gene ID
85463
Gene Name
zinc finger CCCH-type containing 12C
Gene Aliases
MCPIP3
UniGene
Hs.376289
Gene Chromosome Location
Chr.11: 110092425-110171841 on Build GRCh38
Transcripts
NM_033390.1 XM_005271715.3 XM_011543055.2 XM_017018477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627763 c.559G>A p.G187S Chr.11: 110137200-110137200 on Build GRCh38
1627764 c.582T>G p.D194E Chr.11: 110137223-110137223 on Build GRCh38
4165645 c.468T>A p.P156P Chr.11: 110137109-110137109 on Build GRCh38
278189 c.511A>G p.K171E Chr.11: 110137152-110137152 on Build GRCh38
1561103 c.622G>A p.A208T Chr.11: 110137263-110137263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766839736 Chr.11: 110137157-110137157 on Build GRCh38
rs779805436 Chr.11: 110137235-110137235 on Build GRCh38
rs745409793 Chr.11: 110137214-110137214 on Build GRCh38
rs750555920 Chr.11: 110137132-110137132 on Build GRCh38
rs1017125654 Chr.11: 110137197-110137197 on Build GRCh38
rs78240010 Chr.11: 110137109-110137109 on Build GRCh38
rs927143420 Chr.11: 110137126-110137126 on Build GRCh38
rs373888883 Chr.11: 110137274-110137274 on Build GRCh38
rs756984214 Chr.11: 110137202-110137202 on Build GRCh38
rs753475922 Chr.11: 110137187-110137187 on Build GRCh38
rs992678639 Chr.11: 110137190-110137190 on Build GRCh38
rs370181094 Chr.11: 110137116-110137116 on Build GRCh38
rs755252830 Chr.11: 110137172-110137172 on Build GRCh38
rs751932671 Chr.11: 110137168-110137168 on Build GRCh38
rs561816640 Chr.11: 110137270-110137270 on Build GRCh38
rs916962484 Chr.11: 110137191-110137191 on Build GRCh38
rs778554127 Chr.11: 110137203-110137203 on Build GRCh38
rs141278977 Chr.11: 110137124-110137124 on Build GRCh38
rs746997224 Chr.11: 110137239-110137239 on Build GRCh38
rs542273465 Chr.11: 110137280-110137280 on Build GRCh38
rs200420973 Chr.11: 110137110-110137110 on Build GRCh38
rs573118259 Chr.11: 110137148-110137148 on Build GRCh38
rs145474029 Chr.11: 110137182-110137182 on Build GRCh38
rs762024976 Chr.11: 110137120-110137120 on Build GRCh38
rs768266860 Chr.11: 110137247-110137247 on Build GRCh38
rs201652254 Chr.11: 110137273-110137273 on Build GRCh38
rs772024289 Chr.11: 110137220-110137220 on Build GRCh38

View Full Product Details