Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74909477-74909726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KL1
Entrez Gene ID
83694
Gene Name
ribosomal protein S6 kinase like 1
Gene Aliases
RSKL2
UniGene
Hs.414481
Gene Chromosome Location
Chr.14: 74903954-74923396 on Build GRCh38
Transcripts
NM_031464.4 XM_011537206.2 XM_011537207.2 XM_011537211.2 XM_011537213.2 XM_017021680.1 XM_017021681.1 XM_017021682.1 XM_017021683.1 XM_017021684.1 XM_017021685.1 XM_017021686.1 XM_017021687.1 XM_017021688.1 XM_017021689.1 XM_017021690.1 XM_017021691.1 XM_017021692.1

SNP Detail

SNP ID Chromosome Location
rs148616967 Chr.14: 74909529-74909529 on Build GRCh38
rs769196458 Chr.14: 74909526-74909526 on Build GRCh38
rs757893795 Chr.14: 74909574-74909574 on Build GRCh38
rs200092346 Chr.14: 74909568-74909568 on Build GRCh38
rs150539901 Chr.14: 74909597-74909597 on Build GRCh38
rs756024731 Chr.14: 74909693-74909693 on Build GRCh38
rs750796538 Chr.14: 74909559-74909559 on Build GRCh38
rs373499816 Chr.14: 74909514-74909514 on Build GRCh38
rs909443108 Chr.14: 74909705-74909705 on Build GRCh38
rs750232007 Chr.14: 74909690-74909690 on Build GRCh38
rs138644443 Chr.14: 74909601-74909601 on Build GRCh38
rs749418985 Chr.14: 74909702-74909702 on Build GRCh38
rs369994026 Chr.14: 74909571-74909571 on Build GRCh38
rs374560120 Chr.14: 74909576-74909576 on Build GRCh38
rs772568347 Chr.14: 74909617-74909617 on Build GRCh38
rs374989083 Chr.14: 74909654-74909654 on Build GRCh38
rs752409276 Chr.14: 74909562-74909562 on Build GRCh38
rs544708374 Chr.14: 74909510-74909510 on Build GRCh38
rs762699814 Chr.14: 74909639-74909639 on Build GRCh38
rs922227882 Chr.14: 74909683-74909683 on Build GRCh38
rs1001400689 Chr.14: 74909557-74909557 on Build GRCh38
rs773560995 Chr.14: 74909626-74909626 on Build GRCh38
rs764846440 Chr.14: 74909634-74909634 on Build GRCh38
rs957412045 Chr.14: 74909581-74909581 on Build GRCh38
rs972440109 Chr.14: 74909671-74909671 on Build GRCh38
rs752334065 Chr.14: 74909635-74909635 on Build GRCh38
rs777628151 Chr.14: 74909498-74909498 on Build GRCh38
rs200173664 Chr.14: 74909608-74909608 on Build GRCh38
rs754755884 Chr.14: 74909666-74909667 on Build GRCh38
rs575292252 Chr.14: 74909555-74909555 on Build GRCh38
rs775649015 Chr.14: 74909521-74909521 on Build GRCh38
rs759120913 Chr.14: 74909567-74909567 on Build GRCh38
rs56358343 Chr.14: 74909694-74909694 on Build GRCh38
rs770140354 Chr.14: 74909596-74909596 on Build GRCh38
rs993887595 Chr.14: 74909525-74909525 on Build GRCh38
rs751094126 Chr.14: 74909661-74909661 on Build GRCh38
rs770674670 Chr.14: 74909509-74909509 on Build GRCh38
rs758348241 Chr.14: 74909521-74909522 on Build GRCh38
rs370255445 Chr.14: 74909594-74909594 on Build GRCh38
rs369432035 Chr.14: 74909500-74909500 on Build GRCh38
rs749369105 Chr.14: 74909600-74909600 on Build GRCh38
rs201677862 Chr.14: 74909582-74909582 on Build GRCh38
rs763012067 Chr.14: 74909523-74909523 on Build GRCh38
rs757078900 Chr.14: 74909689-74909689 on Build GRCh38
rs779911662 Chr.14: 74909557-74909559 on Build GRCh38
rs751558823 Chr.14: 74909655-74909656 on Build GRCh38
rs752187949 Chr.14: 74909573-74909573 on Build GRCh38
rs781076479 Chr.14: 74909589-74909589 on Build GRCh38
rs761085125 Chr.14: 74909628-74909628 on Build GRCh38
rs762411498 Chr.14: 74909552-74909552 on Build GRCh38
rs774509049 Chr.14: 74909607-74909607 on Build GRCh38
rs932003826 Chr.14: 74909643-74909643 on Build GRCh38
rs55860986 Chr.14: 74909642-74909642 on Build GRCh38

View Full Product Details