Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32311662-32311919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAR1L
Entrez Gene ID
646799
Gene Name
zygote arrest 1-like
Gene Aliases
Z3CXXC7, ZAR2
UniGene
Hs.569254
Gene Chromosome Location
Chr.13: 32303771-32311954 on Build GRCh38
Transcripts
NM_001136571.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1366373 c.197C>T p.A66V Chr.13: 32311729-32311729 on Build GRCh38
4804818 c.261C>A p.N87K Chr.13: 32311665-32311665 on Build GRCh38
4480555 c.242C>T p.P81L Chr.13: 32311684-32311684 on Build GRCh38
7398163 c.149T>C p.L50P Chr.13: 32311777-32311777 on Build GRCh38
1477191 c.139C>G p.P47A Chr.13: 32311787-32311787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991737986 Chr.13: 32311884-32311884 on Build GRCh38
rs560878294 Chr.13: 32311871-32311871 on Build GRCh38
rs752969196 Chr.13: 32311684-32311684 on Build GRCh38
rs983031599 Chr.13: 32311761-32311761 on Build GRCh38
rs758842310 Chr.13: 32311747-32311747 on Build GRCh38
rs563459730 Chr.13: 32311681-32311681 on Build GRCh38
rs868357946 Chr.13: 32311768-32311768 on Build GRCh38
rs948811985 Chr.13: 32311810-32311810 on Build GRCh38
rs761419640 Chr.13: 32311871-32311871 on Build GRCh38
rs925697365 Chr.13: 32311821-32311821 on Build GRCh38
rs530659850 Chr.13: 32311682-32311682 on Build GRCh38
rs967047373 Chr.13: 32311729-32311729 on Build GRCh38
rs758632750 Chr.13: 32311718-32311718 on Build GRCh38
rs768493650 Chr.13: 32311869-32311869 on Build GRCh38
rs908553247 Chr.13: 32311769-32311769 on Build GRCh38
rs749099968 Chr.13: 32311829-32311829 on Build GRCh38
rs915811918 Chr.13: 32311890-32311890 on Build GRCh38
rs751651655 Chr.13: 32311765-32311765 on Build GRCh38
rs780023491 Chr.13: 32311802-32311802 on Build GRCh38
rs991373681 Chr.13: 32311857-32311857 on Build GRCh38
rs938650038 Chr.13: 32311782-32311782 on Build GRCh38
rs950022307 Chr.13: 32311900-32311900 on Build GRCh38
rs778018735 Chr.13: 32311728-32311728 on Build GRCh38
rs778212987 Chr.13: 32311865-32311865 on Build GRCh38
rs903656091 Chr.13: 32311889-32311889 on Build GRCh38
rs935800818 Chr.13: 32311840-32311840 on Build GRCh38
rs992780692 Chr.13: 32311785-32311785 on Build GRCh38
rs542906029 Chr.13: 32311792-32311792 on Build GRCh38
rs75257539 Chr.13: 32311893-32311893 on Build GRCh38

View Full Product Details