Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155910102-155910337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDO2
Entrez Gene ID
6999
Gene Name
tryptophan 2,3-dioxygenase
Gene Aliases
TDO, TO, TPH2, TRPO
UniGene
Hs.183671
Gene Chromosome Location
Chr.4: 155903693-155920406 on Build GRCh38
Transcripts
NM_005651.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6011673 c.618+4T>C p.? Chr.4: 155910215-155910215 on Build GRCh38
1428204 c.513G>A p.M171I Chr.4: 155910106-155910106 on Build GRCh38
5558907 c.612A>G p.L204L Chr.4: 155910205-155910205 on Build GRCh38
6487770 c.599C>A p.T200K Chr.4: 155910192-155910192 on Build GRCh38
1052546 c.585T>C p.S195S Chr.4: 155910178-155910178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765510525 Chr.4: 155910213-155910213 on Build GRCh38
rs761217656 Chr.4: 155910199-155910199 on Build GRCh38
rs780053027 Chr.4: 155910231-155910231 on Build GRCh38
rs754331032 Chr.4: 155910129-155910131 on Build GRCh38
rs1038199774 Chr.4: 155910136-155910136 on Build GRCh38
rs745569234 Chr.4: 155910252-155910252 on Build GRCh38
rs757810917 Chr.4: 155910251-155910253 on Build GRCh38
rs374938550 Chr.4: 155910135-155910135 on Build GRCh38
rs775054654 Chr.4: 155910190-155910190 on Build GRCh38
rs10517626 Chr.4: 155910292-155910292 on Build GRCh38
rs749108868 Chr.4: 155910243-155910243 on Build GRCh38
rs565780468 Chr.4: 155910172-155910172 on Build GRCh38
rs955052353 Chr.4: 155910198-155910198 on Build GRCh38
rs552139246 Chr.4: 155910132-155910132 on Build GRCh38
rs770638838 Chr.4: 155910178-155910178 on Build GRCh38
rs754228949 Chr.4: 155910204-155910204 on Build GRCh38
rs770570430 Chr.4: 155910245-155910245 on Build GRCh38
rs376666725 Chr.4: 155910239-155910239 on Build GRCh38
rs979140988 Chr.4: 155910299-155910299 on Build GRCh38
rs777805051 Chr.4: 155910241-155910241 on Build GRCh38
rs980049667 Chr.4: 155910254-155910254 on Build GRCh38
rs751661404 Chr.4: 155910236-155910236 on Build GRCh38
rs773821631 Chr.4: 155910181-155910181 on Build GRCh38
rs777751949 Chr.4: 155910163-155910163 on Build GRCh38
rs138036577 Chr.4: 155910147-155910147 on Build GRCh38
rs762113400 Chr.4: 155910210-155910210 on Build GRCh38
rs201522196 Chr.4: 155910226-155910226 on Build GRCh38
rs759094500 Chr.4: 155910184-155910184 on Build GRCh38
rs778768261 Chr.4: 155910247-155910247 on Build GRCh38
rs200816290 Chr.4: 155910150-155910150 on Build GRCh38
rs186649136 Chr.4: 155910291-155910291 on Build GRCh38
rs764550228 Chr.4: 155910200-155910200 on Build GRCh38
rs758641246 Chr.4: 155910230-155910230 on Build GRCh38

View Full Product Details