Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70426446-70426679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD2
Entrez Gene ID
54857
Gene Name
glycerophosphodiester phosphodiesterase domain containing 2
Gene Aliases
GDE3, OBDPF
UniGene
Hs.438712
Gene Chromosome Location
Chr.X: 70423031-70433391 on Build GRCh38
Transcripts
NM_001171191.1 NM_001171192.1 NM_001171193.1 NM_017711.3 XM_011530977.1 XM_017029614.1

SNP Detail

SNP ID Chromosome Location
rs764611851 Chr.X: 70426640-70426640 on Build GRCh38
rs906965267 Chr.X: 70426562-70426562 on Build GRCh38
rs779427335 Chr.X: 70426599-70426599 on Build GRCh38
rs770524254 Chr.X: 70426622-70426622 on Build GRCh38
rs1009762845 Chr.X: 70426558-70426558 on Build GRCh38
rs375646779 Chr.X: 70426490-70426490 on Build GRCh38
rs990797016 Chr.X: 70426542-70426542 on Build GRCh38
rs745602670 Chr.X: 70426625-70426625 on Build GRCh38
rs1034019390 Chr.X: 70426621-70426621 on Build GRCh38
rs775617807 Chr.X: 70426497-70426497 on Build GRCh38
rs892699218 Chr.X: 70426523-70426523 on Build GRCh38
rs1046578900 Chr.X: 70426560-70426560 on Build GRCh38
rs769578225 Chr.X: 70426656-70426656 on Build GRCh38
rs369984051 Chr.X: 70426513-70426513 on Build GRCh38
rs756427350 Chr.X: 70426487-70426487 on Build GRCh38
rs200660900 Chr.X: 70426600-70426600 on Build GRCh38

View Full Product Details