Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62566967-62567192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1G
Entrez Gene ID
1937
Gene Name
eukaryotic translation elongation factor 1 gamma
Gene Aliases
EF1G, GIG35
UniGene
Hs.144835
Gene Chromosome Location
Chr.11: 62559601-62573988 on Build GRCh38
Transcripts
NM_001404.4 NR_106805.1

Gene Symbol
MIR6747
Entrez Gene ID
102465447
Gene Name
microRNA 6747
Gene Aliases
hsa-mir-6747
UniGene
Gene Chromosome Location
Chr.11: 62567011-62567071 on Build GRCh38
Transcripts
NM_001404.4 NR_106805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250957 c.695G>A p.R232Q Chr.11: 62566968-62566968 on Build GRCh38
7194678 c.661T>C p.F221L Chr.11: 62567002-62567002 on Build GRCh38
7194679 c.811T>C p.F271L Chr.11: 62567002-62567002 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765288776 Chr.11: 62567004-62567004 on Build GRCh38
rs965251539 Chr.11: 62567145-62567145 on Build GRCh38
rs777444689 Chr.11: 62567097-62567097 on Build GRCh38
rs764504754 Chr.11: 62567067-62567068 on Build GRCh38
rs752673085 Chr.11: 62567146-62567146 on Build GRCh38
rs1016804819 Chr.11: 62567074-62567074 on Build GRCh38
rs769654893 Chr.11: 62567112-62567112 on Build GRCh38
rs770740954 Chr.11: 62567101-62567101 on Build GRCh38
rs371597439 Chr.11: 62566988-62566988 on Build GRCh38
rs758007757 Chr.11: 62567092-62567092 on Build GRCh38
rs955278849 Chr.11: 62567163-62567163 on Build GRCh38
rs369554491 Chr.11: 62567041-62567041 on Build GRCh38
rs763576289 Chr.11: 62567066-62567067 on Build GRCh38
rs918276591 Chr.11: 62567122-62567122 on Build GRCh38
rs11545771 Chr.11: 62566990-62566990 on Build GRCh38
rs959243134 Chr.11: 62567044-62567044 on Build GRCh38
rs778560083 Chr.11: 62567090-62567090 on Build GRCh38
rs776348980 Chr.11: 62567106-62567106 on Build GRCh38
rs986582508 Chr.11: 62567168-62567168 on Build GRCh38
rs376546250 Chr.11: 62567040-62567040 on Build GRCh38
rs1001090242 Chr.11: 62567094-62567094 on Build GRCh38
rs11549631 Chr.11: 62566992-62566992 on Build GRCh38
rs36126146 Chr.11: 62567112-62567113 on Build GRCh38
rs764241530 Chr.11: 62567031-62567031 on Build GRCh38
rs751750965 Chr.11: 62567037-62567037 on Build GRCh38
rs11545770 Chr.11: 62566997-62566997 on Build GRCh38
rs746685016 Chr.11: 62567099-62567099 on Build GRCh38
rs763026799 Chr.11: 62567026-62567026 on Build GRCh38
rs774782969 Chr.11: 62567069-62567070 on Build GRCh38
rs757449375 Chr.11: 62567169-62567169 on Build GRCh38
rs907260700 Chr.11: 62567016-62567016 on Build GRCh38
rs370099895 Chr.11: 62567039-62567039 on Build GRCh38
rs765319189 Chr.11: 62567109-62567109 on Build GRCh38
rs537835649 Chr.11: 62567058-62567058 on Build GRCh38
rs756738855 Chr.11: 62567067-62567067 on Build GRCh38
rs747592809 Chr.11: 62567091-62567091 on Build GRCh38
rs147628715 Chr.11: 62567017-62567017 on Build GRCh38
rs865829861 Chr.11: 62566998-62566998 on Build GRCh38

View Full Product Details