Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 104490742-104490995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXXC4
Entrez Gene ID
80319
Gene Name
CXXC finger protein 4
Gene Aliases
IDAX
UniGene
Hs.12248
Gene Chromosome Location
Chr.4: 104468306-104494967 on Build GRCh38
Transcripts
NM_025212.3 NR_132741.1 XM_011532284.2 XM_017008652.1 NR_125926.1

Gene Symbol
LOC101929468
Entrez Gene ID
101929468
Gene Name
uncharacterized LOC101929468
Gene Aliases
-
UniGene
Hs.480435
Gene Chromosome Location
Chr.4: 104490965-104697592 on Build GRCh38
Transcripts
NM_025212.3 NR_132741.1 XM_011532284.2 XM_017008652.1 NR_125926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588937 c.552+1G>C p.? Chr.4: 104490743-104490743 on Build GRCh38
3598840 c.986G>A p.R329H Chr.4: 104490817-104490817 on Build GRCh38
2952580 c.369A>G p.S123S Chr.4: 104490927-104490927 on Build GRCh38
3974645 c.878G>A p.G293E Chr.4: 104490925-104490925 on Build GRCh38
6638698 c.528G>T p.K176N Chr.4: 104490768-104490768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746953946 Chr.4: 104490786-104490786 on Build GRCh38
rs774945135 Chr.4: 104490861-104490861 on Build GRCh38
rs779928606 Chr.4: 104490771-104490771 on Build GRCh38
rs370500943 Chr.4: 104490920-104490920 on Build GRCh38
rs781229081 Chr.4: 104490948-104490948 on Build GRCh38
rs372212419 Chr.4: 104490942-104490942 on Build GRCh38
rs750088127 Chr.4: 104490937-104490939 on Build GRCh38
rs762670643 Chr.4: 104490929-104490929 on Build GRCh38
rs556281831 Chr.4: 104490810-104490810 on Build GRCh38
rs199602357 Chr.4: 104490962-104490962 on Build GRCh38
rs768141659 Chr.4: 104490902-104490902 on Build GRCh38
rs758211749 Chr.4: 104490768-104490768 on Build GRCh38
rs754865850 Chr.4: 104490943-104490943 on Build GRCh38
rs555311957 Chr.4: 104490940-104490940 on Build GRCh38
rs761593062 Chr.4: 104490937-104490945 on Build GRCh38
rs748327160 Chr.4: 104490817-104490817 on Build GRCh38
rs571960913 Chr.4: 104490960-104490960 on Build GRCh38
rs146939312 Chr.4: 104490939-104490939 on Build GRCh38
rs147556012 Chr.4: 104490959-104490959 on Build GRCh38
rs377103900 Chr.4: 104490903-104490903 on Build GRCh38
rs764745722 Chr.4: 104490936-104490937 on Build GRCh38
rs750108778 Chr.4: 104490922-104490922 on Build GRCh38
rs760109133 Chr.4: 104490873-104490873 on Build GRCh38
rs867889302 Chr.4: 104490930-104490930 on Build GRCh38
rs772336462 Chr.4: 104490927-104490927 on Build GRCh38
rs189558755 Chr.4: 104490832-104490832 on Build GRCh38
rs185047970 Chr.4: 104490822-104490822 on Build GRCh38
rs755026903 Chr.4: 104490789-104490789 on Build GRCh38
rs965597760 Chr.4: 104490869-104490869 on Build GRCh38
rs761436810 Chr.4: 104490904-104490904 on Build GRCh38
rs766173397 Chr.4: 104490937-104490937 on Build GRCh38
rs751412118 Chr.4: 104490938-104490938 on Build GRCh38
rs771450633 Chr.4: 104490846-104490846 on Build GRCh38
rs746334700 Chr.4: 104490840-104490840 on Build GRCh38

View Full Product Details