Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 853153-853394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELANE
Entrez Gene ID
1991
Gene Name
elastase, neutrophil expressed
Gene Aliases
ELA2, GE, HLE, HNE, NE, PMN-E, SCN1
UniGene
Hs.99863
Gene Chromosome Location
Chr.19: 850997-856250 on Build GRCh38
Transcripts
NM_001972.3 XM_011527775.1 XM_011527776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680698 c.287G>T p.R96L Chr.19: 853324-853324 on Build GRCh38
4680699 c.322G>A p.G108S Chr.19: 853359-853359 on Build GRCh38
1238773 c.301G>C p.V101L Chr.19: 853338-853338 on Build GRCh38
5857159 c.269C>T p.S90L Chr.19: 853306-853306 on Build GRCh38
3541611 c.231C>T p.V77V Chr.19: 853268-853268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200627242 Chr.19: 853334-853334 on Build GRCh38
rs758933736 Chr.19: 853214-853214 on Build GRCh38
rs777599059 Chr.19: 853256-853256 on Build GRCh38
rs751974314 Chr.19: 853220-853220 on Build GRCh38
rs899133377 Chr.19: 853299-853299 on Build GRCh38
rs769123461 Chr.19: 853272-853272 on Build GRCh38
rs137854449 Chr.19: 853338-853338 on Build GRCh38
rs780393819 Chr.19: 853233-853233 on Build GRCh38
rs981872389 Chr.19: 853222-853222 on Build GRCh38
rs351105 Chr.19: 853234-853234 on Build GRCh38
rs1033255773 Chr.19: 853179-853179 on Build GRCh38
rs191587 Chr.19: 853208-853208 on Build GRCh38
rs745455816 Chr.19: 853345-853345 on Build GRCh38
rs755088297 Chr.19: 853312-853312 on Build GRCh38
rs200503891 Chr.19: 853257-853257 on Build GRCh38
rs761938801 Chr.19: 853257-853257 on Build GRCh38
rs752760119 Chr.19: 853321-853321 on Build GRCh38
rs267606781 Chr.19: 853329-853329 on Build GRCh38
rs777652186 Chr.19: 853323-853323 on Build GRCh38
rs771866084 Chr.19: 853254-853254 on Build GRCh38
rs747966670 Chr.19: 853251-853251 on Build GRCh38
rs753965718 Chr.19: 853305-853305 on Build GRCh38
rs767938859 Chr.19: 853224-853224 on Build GRCh38
rs768135268 Chr.19: 853280-853280 on Build GRCh38
rs988374919 Chr.19: 853203-853203 on Build GRCh38
rs754000264 Chr.19: 853236-853236 on Build GRCh38
rs940366307 Chr.19: 853243-853243 on Build GRCh38
rs778939927 Chr.19: 853250-853250 on Build GRCh38
rs890280050 Chr.19: 853306-853306 on Build GRCh38
rs778800388 Chr.19: 853314-853314 on Build GRCh38
rs559254755 Chr.19: 853196-853196 on Build GRCh38
rs369606303 Chr.19: 853371-853371 on Build GRCh38
rs532963460 Chr.19: 853221-853221 on Build GRCh38
rs780905514 Chr.19: 853344-853344 on Build GRCh38
rs143634717 Chr.19: 853296-853296 on Build GRCh38
rs549887145 Chr.19: 853335-853335 on Build GRCh38
rs746799175 Chr.19: 853265-853265 on Build GRCh38
rs200786414 Chr.19: 853358-853358 on Build GRCh38
rs770660263 Chr.19: 853341-853341 on Build GRCh38
rs569608929 Chr.19: 853270-853270 on Build GRCh38
rs996641598 Chr.19: 853367-853367 on Build GRCh38
rs774932781 Chr.19: 853370-853370 on Build GRCh38
rs878855323 Chr.19: 853306-853307 on Build GRCh38
rs536244338 Chr.19: 853217-853217 on Build GRCh38
rs754964823 Chr.19: 853240-853240 on Build GRCh38
rs200002231 Chr.19: 853337-853337 on Build GRCh38
rs949525993 Chr.19: 853215-853215 on Build GRCh38
rs201882944 Chr.19: 853273-853273 on Build GRCh38
rs764707430 Chr.19: 853219-853219 on Build GRCh38
rs766647358 Chr.19: 853301-853301 on Build GRCh38
rs568474579 Chr.19: 853342-853342 on Build GRCh38
rs750859137 Chr.19: 853294-853294 on Build GRCh38
rs1008048630 Chr.19: 853324-853324 on Build GRCh38
rs758393927 Chr.19: 853322-853322 on Build GRCh38
rs756473168 Chr.19: 853230-853230 on Build GRCh38
rs551223811 Chr.19: 853227-853227 on Build GRCh38

View Full Product Details