Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 236166078-236166341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBX2
Entrez Gene ID
2637
Gene Name
gastrulation brain homeobox 2
Gene Aliases
-
UniGene
Hs.184945
Gene Chromosome Location
Chr.2: 236165236-236168009 on Build GRCh38
Transcripts
NM_001301687.1 NM_001485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1253122 c.705C>T p.S235S Chr.2: 236166256-236166256 on Build GRCh38
1326316 c.775C>G p.L259V Chr.2: 236166186-236166186 on Build GRCh38
6188941 c.774G>T p.Q258H Chr.2: 236166187-236166187 on Build GRCh38
1195494 c.847G>A p.A283T Chr.2: 236166114-236166114 on Build GRCh38
4686576 c.652G>A p.A218T Chr.2: 236166309-236166309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763225082 Chr.2: 236166248-236166248 on Build GRCh38
rs771308424 Chr.2: 236166224-236166224 on Build GRCh38
rs1003013457 Chr.2: 236166177-236166177 on Build GRCh38
rs778580912 Chr.2: 236166282-236166282 on Build GRCh38
rs755708214 Chr.2: 236166134-236166134 on Build GRCh38
rs866290647 Chr.2: 236166135-236166135 on Build GRCh38
rs749898317 Chr.2: 236166132-236166132 on Build GRCh38
rs748875811 Chr.2: 236166157-236166157 on Build GRCh38
rs143663937 Chr.2: 236166296-236166296 on Build GRCh38
rs113681406 Chr.2: 236166256-236166256 on Build GRCh38
rs757092821 Chr.2: 236166126-236166126 on Build GRCh38
rs975760254 Chr.2: 236166272-236166272 on Build GRCh38
rs775661503 Chr.2: 236166310-236166310 on Build GRCh38
rs201407896 Chr.2: 236166229-236166229 on Build GRCh38
rs753044489 Chr.2: 236166207-236166208 on Build GRCh38
rs757696070 Chr.2: 236166290-236166290 on Build GRCh38
rs371662608 Chr.2: 236166142-236166142 on Build GRCh38
rs7588069 Chr.2: 236166184-236166184 on Build GRCh38
rs769851621 Chr.2: 236166236-236166236 on Build GRCh38
rs754607396 Chr.2: 236166277-236166277 on Build GRCh38
rs767344478 Chr.2: 236166257-236166257 on Build GRCh38
rs764433841 Chr.2: 236166251-236166251 on Build GRCh38
rs747458810 Chr.2: 236166216-236166216 on Build GRCh38
rs372760556 Chr.2: 236166214-236166214 on Build GRCh38
rs369589992 Chr.2: 236166100-236166100 on Build GRCh38
rs781421148 Chr.2: 236166291-236166291 on Build GRCh38
rs373486610 Chr.2: 236166260-236166260 on Build GRCh38
rs537846896 Chr.2: 236166108-236166108 on Build GRCh38
rs779554048 Chr.2: 236166136-236166136 on Build GRCh38
rs201674356 Chr.2: 236166252-236166252 on Build GRCh38
rs749427581 Chr.2: 236166317-236166317 on Build GRCh38
rs746318204 Chr.2: 236166295-236166295 on Build GRCh38
rs202152349 Chr.2: 236166276-236166276 on Build GRCh38
rs200791522 Chr.2: 236166230-236166230 on Build GRCh38
rs763280325 Chr.2: 236166107-236166107 on Build GRCh38
rs921209289 Chr.2: 236166314-236166314 on Build GRCh38
rs781231207 Chr.2: 236166127-236166127 on Build GRCh38
rs376571505 Chr.2: 236166247-236166247 on Build GRCh38
rs751429932 Chr.2: 236166116-236166116 on Build GRCh38
rs988304642 Chr.2: 236166183-236166183 on Build GRCh38
rs745919266 Chr.2: 236166233-236166233 on Build GRCh38
rs756011584 Chr.2: 236166268-236166268 on Build GRCh38
rs747326842 Chr.2: 236166289-236166289 on Build GRCh38

View Full Product Details