Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BRAF
- Entrez Gene ID
-
673
- Gene Name
- B-Raf proto-oncogene, serine/threonine kinase
- Gene Aliases
-
B-RAF1, B-raf, BRAF1, NS7, RAFB1
- Chromosome Location
-
Chr.7: 140719331 - 140924764 on Build GRCh38
- Gene Symbol
- KIAA1549
- Entrez Gene ID
-
57670
- Gene Name
- KIAA1549
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 138831377 - 138981626 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AM989477.1 |
- |
- |
1898 |
118 |
View Full Product Details