Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156595095-156595352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH4
Entrez Gene ID
54865
Gene Name
G-patch domain containing 4
Gene Aliases
GPATC4
UniGene
Hs.193832
Gene Chromosome Location
Chr.1: 156594308-156601496 on Build GRCh38
Transcripts
NM_015590.3 NM_182679.2 XM_005245287.3 XM_017000319.1

Gene Symbol
NAXE
Entrez Gene ID
128240
Gene Name
NAD(P)HX epimerase
Gene Aliases
AIBP, APOA1BP, YJEFN1
UniGene
Hs.528320
Gene Chromosome Location
Chr.1: 156591762-156599818 on Build GRCh38
Transcripts
NM_015590.3 NM_182679.2 XM_005245287.3 XM_017000319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4222795 c.1087A>T p.R363W Chr.1: 156595254-156595254 on Build GRCh38
6990617 c.1050G>C p.E350D Chr.1: 156595291-156595291 on Build GRCh38
3376756 c.1046A>C p.E349A Chr.1: 156595295-156595295 on Build GRCh38
6990618 c.1035G>C p.E345D Chr.1: 156595291-156595291 on Build GRCh38
5022132 c.1083_1084insGT p.F362fs*>15 Chr.1: 156595257-156595258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77528517 Chr.1: 156595257-156595257 on Build GRCh38
rs1016632736 Chr.1: 156595154-156595154 on Build GRCh38
rs752767548 Chr.1: 156595313-156595316 on Build GRCh38
rs190829263 Chr.1: 156595127-156595127 on Build GRCh38
rs755732138 Chr.1: 156595285-156595285 on Build GRCh38
rs374578579 Chr.1: 156595243-156595243 on Build GRCh38
rs62641592 Chr.1: 156595206-156595206 on Build GRCh38
rs202077240 Chr.1: 156595120-156595120 on Build GRCh38
rs201712425 Chr.1: 156595319-156595319 on Build GRCh38
rs780754846 Chr.1: 156595141-156595141 on Build GRCh38
rs762993333 Chr.1: 156595221-156595221 on Build GRCh38
rs770651900 Chr.1: 156595217-156595220 on Build GRCh38
rs762116911 Chr.1: 156595204-156595204 on Build GRCh38
rs762756481 Chr.1: 156595204-156595206 on Build GRCh38
rs1018229987 Chr.1: 156595242-156595242 on Build GRCh38
rs3795733 Chr.1: 156595130-156595130 on Build GRCh38
rs368411369 Chr.1: 156595271-156595271 on Build GRCh38
rs745815820 Chr.1: 156595325-156595325 on Build GRCh38
rs72559129 Chr.1: 156595257-156595258 on Build GRCh38
rs10637036 Chr.1: 156595258-156595259 on Build GRCh38
rs778845960 Chr.1: 156595323-156595323 on Build GRCh38
rs777336317 Chr.1: 156595290-156595290 on Build GRCh38
rs138933759 Chr.1: 156595322-156595322 on Build GRCh38
rs142485230 Chr.1: 156595187-156595187 on Build GRCh38
rs766863626 Chr.1: 156595230-156595230 on Build GRCh38
rs770875628 Chr.1: 156595180-156595180 on Build GRCh38
rs368315395 Chr.1: 156595195-156595195 on Build GRCh38
rs866563657 Chr.1: 156595256-156595257 on Build GRCh38
rs772198575 Chr.1: 156595332-156595332 on Build GRCh38
rs747810197 Chr.1: 156595155-156595155 on Build GRCh38
rs755822109 Chr.1: 156595165-156595165 on Build GRCh38
rs759835961 Chr.1: 156595262-156595262 on Build GRCh38
rs767765471 Chr.1: 156595289-156595291 on Build GRCh38
rs766129211 Chr.1: 156595158-156595158 on Build GRCh38
rs748784417 Chr.1: 156595167-156595167 on Build GRCh38
rs752039712 Chr.1: 156595252-156595252 on Build GRCh38
rs141105131 Chr.1: 156595218-156595218 on Build GRCh38
rs370360354 Chr.1: 156595186-156595186 on Build GRCh38
rs753126379 Chr.1: 156595282-156595282 on Build GRCh38
rs769661827 Chr.1: 156595184-156595184 on Build GRCh38
rs747675457 Chr.1: 156595180-156595181 on Build GRCh38
rs773096395 Chr.1: 156595193-156595194 on Build GRCh38
rs772000500 Chr.1: 156595192-156595192 on Build GRCh38
rs780629224 Chr.1: 156595159-156595161 on Build GRCh38
rs777331336 Chr.1: 156595166-156595166 on Build GRCh38
rs75463812 Chr.1: 156595259-156595259 on Build GRCh38

View Full Product Details