Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 154242579-154242820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT13
Entrez Gene ID
114805
Gene Name
polypeptide N-acetylgalactosaminyltransferase 13
Gene Aliases
GalNAc-T13
UniGene
Hs.470277
Gene Chromosome Location
Chr.2: 153871898-154454177 on Build GRCh38
Transcripts
NM_001301627.1 NM_052917.3 XM_011510537.2 XM_011510538.2 XM_017003258.1 XM_017003259.1 XM_017003260.1 XM_017003261.1 XM_017003262.1 XM_017003263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4085711 c.526C>T p.P176S Chr.2: 154242745-154242745 on Build GRCh38
5058163 c.600A>G p.K200K Chr.2: 154242819-154242819 on Build GRCh38
3299283 c.583G>A p.G195R Chr.2: 154242802-154242802 on Build GRCh38
570304 c.581G>A p.R194Q Chr.2: 154242800-154242800 on Build GRCh38
5091052 c.534A>C p.K178N Chr.2: 154242753-154242753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757930080 Chr.2: 154242795-154242795 on Build GRCh38
rs201676227 Chr.2: 154242684-154242684 on Build GRCh38
rs750465662 Chr.2: 154242647-154242647 on Build GRCh38
rs374708934 Chr.2: 154242714-154242714 on Build GRCh38
rs142707218 Chr.2: 154242729-154242729 on Build GRCh38
rs374003992 Chr.2: 154242800-154242800 on Build GRCh38
rs540619836 Chr.2: 154242666-154242666 on Build GRCh38
rs907197681 Chr.2: 154242702-154242702 on Build GRCh38
rs138158527 Chr.2: 154242754-154242754 on Build GRCh38
rs946257841 Chr.2: 154242682-154242682 on Build GRCh38
rs776718504 Chr.2: 154242727-154242727 on Build GRCh38
rs777417959 Chr.2: 154242676-154242676 on Build GRCh38
rs537023620 Chr.2: 154242675-154242675 on Build GRCh38
rs1055008406 Chr.2: 154242658-154242658 on Build GRCh38
rs187976407 Chr.2: 154242726-154242726 on Build GRCh38
rs1018857509 Chr.2: 154242799-154242799 on Build GRCh38
rs9288324 Chr.2: 154242633-154242633 on Build GRCh38
rs757677744 Chr.2: 154242673-154242673 on Build GRCh38
rs752527151 Chr.2: 154242667-154242667 on Build GRCh38
rs745351247 Chr.2: 154242686-154242686 on Build GRCh38
rs753565309 Chr.2: 154242773-154242773 on Build GRCh38
rs938836094 Chr.2: 154242611-154242611 on Build GRCh38
rs368498154 Chr.2: 154242685-154242685 on Build GRCh38
rs748905593 Chr.2: 154242677-154242677 on Build GRCh38
rs772749008 Chr.2: 154242690-154242690 on Build GRCh38
rs898555268 Chr.2: 154242747-154242747 on Build GRCh38
rs758615923 Chr.2: 154242654-154242654 on Build GRCh38
rs750564424 Chr.2: 154242742-154242742 on Build GRCh38
rs935411215 Chr.2: 154242699-154242699 on Build GRCh38
rs778401789 Chr.2: 154242788-154242788 on Build GRCh38
rs761139906 Chr.2: 154242720-154242720 on Build GRCh38
rs149581963 Chr.2: 154242757-154242757 on Build GRCh38
rs759879455 Chr.2: 154242787-154242787 on Build GRCh38
rs752574420 Chr.2: 154242763-154242763 on Build GRCh38
rs532958580 Chr.2: 154242608-154242609 on Build GRCh38
rs765363190 Chr.2: 154242736-154242736 on Build GRCh38
rs777690153 Chr.2: 154242772-154242772 on Build GRCh38
rs780716644 Chr.2: 154242797-154242797 on Build GRCh38
rs745417238 Chr.2: 154242794-154242794 on Build GRCh38
rs755908617 Chr.2: 154242768-154242768 on Build GRCh38
rs762987864 Chr.2: 154242746-154242746 on Build GRCh38

View Full Product Details