Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 20705876-20706145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLCO1C1
Entrez Gene ID
53919
Gene Name
solute carrier organic anion transporter family member 1C1
Gene Aliases
OATP-F, OATP1, OATP14, OATP1C1, OATPF, OATPRP5, SLC21A14
UniGene
Hs.47261
Gene Chromosome Location
Chr.12: 20690505-20753386 on Build GRCh38
Transcripts
NM_001145944.1 NM_001145945.1 NM_001145946.1 NM_017435.4 XM_005253394.2 XM_005253396.2 XM_011520703.2 XM_011520704.2 XM_011520709.2 XM_011520711.2 XM_017019484.1 XM_017019485.1 XM_017019486.1 XM_017019487.1 XM_017019488.1 XM_017019489.1 XM_017019490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1266099 c.394T>A p.F132I Chr.12: 20706071-20706071 on Build GRCh38
692877 c.272-1G>A p.? Chr.12: 20705948-20705948 on Build GRCh38
315402 c.354C>A p.I118I Chr.12: 20706031-20706031 on Build GRCh38
5667414 c.311C>A p.A104D Chr.12: 20705988-20705988 on Build GRCh38
7412246 c.315A>T p.K105N Chr.12: 20705992-20705992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762848714 Chr.12: 20706004-20706004 on Build GRCh38
rs764049864 Chr.12: 20706017-20706017 on Build GRCh38
rs1040240916 Chr.12: 20705985-20705985 on Build GRCh38
rs772854324 Chr.12: 20705930-20705930 on Build GRCh38
rs570492269 Chr.12: 20706033-20706033 on Build GRCh38
rs201099004 Chr.12: 20706101-20706101 on Build GRCh38
rs1005816700 Chr.12: 20706074-20706074 on Build GRCh38
rs768948685 Chr.12: 20705929-20705929 on Build GRCh38
rs769284258 Chr.12: 20706082-20706082 on Build GRCh38
rs748489307 Chr.12: 20706069-20706069 on Build GRCh38
rs200993713 Chr.12: 20705941-20705941 on Build GRCh38
rs865879669 Chr.12: 20705954-20705954 on Build GRCh38
rs765109613 Chr.12: 20705987-20705987 on Build GRCh38
rs746729301 Chr.12: 20705939-20705939 on Build GRCh38
rs368631619 Chr.12: 20705960-20705960 on Build GRCh38
rs1012342770 Chr.12: 20705977-20705977 on Build GRCh38
rs763045986 Chr.12: 20706099-20706099 on Build GRCh38
rs780566296 Chr.12: 20705924-20705924 on Build GRCh38
rs796840888 Chr.12: 20705921-20705921 on Build GRCh38
rs148984880 Chr.12: 20706076-20706076 on Build GRCh38
rs373359001 Chr.12: 20706047-20706047 on Build GRCh38
rs369951135 Chr.12: 20706107-20706107 on Build GRCh38
rs931919021 Chr.12: 20706022-20706022 on Build GRCh38
rs17852030 Chr.12: 20706052-20706052 on Build GRCh38
rs757424194 Chr.12: 20705927-20705927 on Build GRCh38
rs201650716 Chr.12: 20706038-20706038 on Build GRCh38
rs781191629 Chr.12: 20705907-20705907 on Build GRCh38
rs375035942 Chr.12: 20706058-20706058 on Build GRCh38
rs750496047 Chr.12: 20706035-20706035 on Build GRCh38
rs923094661 Chr.12: 20706000-20706000 on Build GRCh38
rs538277486 Chr.12: 20706027-20706027 on Build GRCh38
rs371332779 Chr.12: 20706075-20706075 on Build GRCh38
rs745384082 Chr.12: 20706077-20706077 on Build GRCh38
rs180791246 Chr.12: 20706117-20706117 on Build GRCh38
rs756166916 Chr.12: 20706046-20706046 on Build GRCh38
rs774332229 Chr.12: 20706102-20706102 on Build GRCh38
rs754273890 Chr.12: 20706049-20706049 on Build GRCh38
rs750474101 Chr.12: 20705984-20705984 on Build GRCh38
rs751389365 Chr.12: 20706024-20706024 on Build GRCh38
rs371858674 Chr.12: 20705996-20705996 on Build GRCh38
rs759047567 Chr.12: 20705963-20705963 on Build GRCh38
rs372692156 Chr.12: 20705914-20705914 on Build GRCh38
rs552667648 Chr.12: 20705923-20705923 on Build GRCh38
rs756285912 Chr.12: 20706001-20706001 on Build GRCh38
rs774861012 Chr.12: 20706094-20706094 on Build GRCh38

View Full Product Details