Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48460658-48460901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A5
Entrez Gene ID
92745
Gene Name
solute carrier family 38 member 5
Gene Aliases
JM24, SN2, SNAT5, pp7194
UniGene
Hs.195155
Gene Chromosome Location
Chr.X: 48458537-48470256 on Build GRCh38
Transcripts
NM_033518.3 XM_005272694.3 XM_005272695.4 XM_005272697.2 XM_005272698.4 XM_006724569.3 XM_017029960.1 XM_017029961.1

SNP Detail

SNP ID Chromosome Location
rs782386875 Chr.X: 48460752-48460752 on Build GRCh38
rs782815472 Chr.X: 48460788-48460788 on Build GRCh38
rs190491934 Chr.X: 48460837-48460837 on Build GRCh38
rs368812725 Chr.X: 48460676-48460676 on Build GRCh38
rs781894215 Chr.X: 48460790-48460790 on Build GRCh38
rs137986694 Chr.X: 48460712-48460712 on Build GRCh38
rs782746661 Chr.X: 48460751-48460751 on Build GRCh38
rs143580047 Chr.X: 48460715-48460715 on Build GRCh38
rs113641537 Chr.X: 48460730-48460730 on Build GRCh38
rs781871991 Chr.X: 48460774-48460774 on Build GRCh38
rs1027234037 Chr.X: 48460706-48460706 on Build GRCh38
rs200111614 Chr.X: 48460682-48460682 on Build GRCh38
rs150316312 Chr.X: 48460694-48460694 on Build GRCh38
rs113694771 Chr.X: 48460738-48460738 on Build GRCh38
rs782210012 Chr.X: 48460688-48460688 on Build GRCh38
rs782383746 Chr.X: 48460693-48460693 on Build GRCh38

View Full Product Details